Skip to Content
Merck
All Photos(1)

Documents

P1918

Sigma-Aldrich

Pancuronium bromide

Synonym(s):

1,1′-[(2β,3α,5α,16β,17β)-3,17-bis(Acetyloxy)androstane-2,16-diyl]bis(1-methylpiperidinium) dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H60Br2N2O4
CAS Number:
Molecular Weight:
732.67
Beilstein:
4226892
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

originator

Organon

SMILES string

[Br-].[Br-].[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@]4(C)[C@@H](OC(C)=O)[C@H](C[C@@]34[H])[N+]5(C)CCCCC5)[C@@]1(C)C[C@@H]([C@H](C2)OC(C)=O)[N+]6(C)CCCCC6

InChI

1S/C35H60N2O4.2BrH/c1-24(38)40-32-21-26-13-14-27-28(35(26,4)23-31(32)37(6)19-11-8-12-20-37)15-16-34(3)29(27)22-30(33(34)41-25(2)39)36(5)17-9-7-10-18-36;;/h26-33H,7-23H2,1-6H3;2*1H/q+2;;/p-2/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;;/m0../s1

InChI key

NPIJXCQZLFKBMV-YTGGZNJNSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pancuronium bromide is a biquarternary amino steroid. It blocks muscarinic acetylcholine receptors (M2) in the heart. Pancuronium has vagolytic and sympathomimetic properties. It is associated with tachycardia and hypertension.

Application

Pancuronium Bromide has been used as an analgesic in various experiments.

Biochem/physiol Actions

Aminosteroidal neuromuscular blocking agent; skeletal muscle relaxant

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Organon. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drain's PeriAnesthesia Nursing - E-Book: A Critical Care Approach (2012)
Alexandria N Hughes et al.
Nature communications, 10(1), 4125-4125 (2019-09-13)
Vesicular release from neurons promotes myelin sheath growth on axons. Oligodendrocytes express proteins that allow dendrites to respond to vesicular release at synapses, suggesting that axon-myelin contacts use similar communication mechanisms as synapses to form myelin sheaths. To test this
Fluid osmolarity acutely and differentially modulates lymphatic vessels intrinsic contractions and lymph flow
Solari E, et al.
Frontiers in physiology, 9, 871-871 (2018)
Smith and Aitkenhead's Textbook of Anaesthesia E-Book: Expert Consult - Online & Print (2013)
Raphaël Candelier et al.
Scientific reports, 5, 12196-12196 (2015-07-22)
Zebrafish larva is a unique model for whole-brain functional imaging and to study sensory-motor integration in the vertebrate brain. To take full advantage of this system, one needs to design sensory environments that can mimic the complex spatiotemporal stimulus patterns

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service