Skip to Content
Merck
All Photos(1)

Key Documents

76904

Sigma-Aldrich

Vecuronium bromide

Synonym(s):

1-[(2β,3α,5α,16β,17β)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-piperidinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H57BrN2O4
CAS Number:
Molecular Weight:
637.73
Beilstein:
4837308
EC Number:
MDL number:
UNSPSC Code:
51152000
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥97.0% (TLC)

form

powder or crystals

functional group

carboxylic acid

SMILES string

[Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(C)=O)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1N6CCCCC6

InChI

1S/C34H57N2O4.BrH/c1-23(37)39-31-20-25-12-13-26-27(34(25,4)22-29(31)35-16-8-6-9-17-35)14-15-33(3)28(26)21-30(32(33)40-24(2)38)36(5)18-10-7-11-19-36;/h25-32H,6-22H2,1-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1

InChI key

VEPSYABRBFXYIB-PWXDFCLTSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Vecuronium Bromide is an aminosteroidal neuromuscular blocking agent. It relaxes the skeletal muscle and decreases oxygen consumption during surgery. It competes with acetylcholine and bind to cholinergic receptors at neuromuscular junctions.[1][2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hong WANG et al.
Acta pharmacologica Sinica, 31(12), 1541-1546 (2010-11-26)
to investigate the changing resistance to nondepolarizing muscle relaxants (NDMRs) during the first month after denervation. the denervated and innervated skeletal muscle cells were examined on days 1, 4, 7, 14, 21, and 28 after denervation. Individual denervated and innervated
Shawn P Vorce et al.
Journal of analytical toxicology, 32(6), 422-427 (2008-07-26)
The apparent recreational use of an aminosteroidal non-depolarizing neuromuscular blocking agent, vecuronium, is reported in this postmortem investigation. A quantitative method for the analysis of vecuronium and its active metabolite, 3-desacetylvecuronium, in blood and tissue samples was developed using liquid
Hong Wang et al.
Acta pharmacologica Sinica, 32(12), 1454-1459 (2011-10-12)
To investigate the effect of magnesium sulfate and its interaction with the non-depolarizing muscle relaxant vecuronium at adult muscle-type acetylcholine receptors in vitro. Adult muscle-type acetylcholine receptors were expressed in HEK293 cells. Drug-containing solution was applied via a gravity-driven perfusion
K R Vega-Villa et al.
British journal of anaesthesia, 112(2), 304-310 (2013-09-27)
Burn patients develop resistance to non-depolarizing neuromuscular blocking agents (NDNMBAs) and require a significantly large dose to produce a desired clinical response. Pathophysiological changes related to burn injury may alter pharmacokinetics (PK) and pharmacodynamics of NDNMBAs. The purpose of this
W C Bowman
British journal of pharmacology, 147 Suppl 1, S277-S286 (2006-01-13)
Descriptions of the South American arrow poisons known as curares were reported by explorers in the 16th century, and their site of action in producing neuromuscular block was determined by Claude Bernard in the mid-19th century. Tubocurarine, the most important

Questions

  1. Please let us know the next retest/Expiry date if it is not mentioned in the COA or bottle given for Vecuronium Bromide, Product Number 76904, Batch Number BCCJ5183, thanks. Or please guide, how to check the same

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service