Skip to Content
Merck

P7545

Pancreatin from porcine pancreas

8 × USP specifications

Synonym(s):

Pancreatin from hog pancreas

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
232-468-9
NACRES:
NA.54
MDL number:
Specific activity:
8 × USP specifications
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

specific activity

8 × USP specifications

contains

lactose as extender

storage temp.

−20°C

Application

The enzyme has been used along with amyloglucosidase for the in vitro digestion of starch in food samples. Pancreatin from Sigma has been used for in vitro pepsin-pancreatin digestion assays while estimating of lower gut digestibility using rumen feed residues. It has also been used along with pepsin to simulate in vitro gastric and ileal digestion of raw materials in pigs.
Pancreatin is a mixture of several digestive enzymes produced by the exocrine cells of the porcine pancreas. It is a broad-spectrum protease composed of amylase, trypsin, lipase, ribonuclease and protease. It is used for in vitro digestibility analysis. Product P7545 has been used to test the sensitivities of cellulolytic bacteria inhibitors.

Biochem/physiol Actions

Due to the enzymatic components such as trypsin, amylase and lipase, pancreatin hydrolyzes proteins, starch and fats. Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40 °C, will digest not less than 25 times its weight of casein in 60 minutes at pH7.5 at 40 °C and will release not less than 2 microequivalents of acid per min per mg pancreatin from olive oil at pH9.0 at 37 °C.
Pancreatin contains enzymatic components including trypsin, amylase and lipase, ribonuclease, and protease, produced by the exocrine cells of the porcine pancreas. This combination of enzymes allows it to hydrolyze proteins, starch and fats. Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40°C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40°C and will release not less than microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37°C.


Still not finding the right product?

Explore all of our products under Pancreatin from porcine pancreas


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library