Skip to Content
Merck

Skip To

B8631

Bile extract porcine

Synonym(s):

Bile acids, Hog Bile Powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-369-0
NACRES:
NA.25
MDL number:
Form:
powder
Biological source:
Porcine
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine

Quality Segment

form

powder

technique(s)

microbiological culture: suitable

Application

Bile extract porcine has been used:
  • as a supplement in brain heart infusion (BHI) medium to culture Listeria monocytogenes LO28[1]
  • to investigate the survival of probiotic bacteria through the gastric-small intestine[2]
  • as a supplement in BHI mediato study the effect of oxygen on bile resistance in Listeria monocytogenes[3]

Chemical derivatization and HPLC have been proven for separation and identification of complex mixtures of bile acids and bile acid conjugates.

Biochem/physiol Actions

Bile from bovine and ovine can be used in bacteriological research to investigate bile salt adaptation of bacteria such as Lactobacillus.

Analysis Note

Contains glycine and taurine conjugates of hyodeoxycholic acid and other bile salts.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G1794B8381P3292
technique(s)

microbiological culture: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

microbiological culture: suitable

technique(s)

cell culture | mammalian: suitable

biological source

Porcine

biological source

human

biological source

bovine

biological source

Porcine pancreas

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. mM 단위로 녹여서 사용하고자 하는데, 해당 제품의 분자량을 알수 있을까요?? 제품/MSDS 등에도 써있지 않아서 문의 남깁니다.

    1 answer
    1. There is no singular molecular weight for this product. Bile extract porcine is the dried fraction from hog bile, which is soluble in 80% alcohol. As estimated by TLC, this product contains Hyodeoxycholic acid at 1-5%, Deoxycholic acid at 0.5-7%, Cholic acid at 0.5-2%, Glycodeoxycholic acid at 10-15%, and Taurodeoxycholic acid at 3-9%.

      Helpful?

  2. What is the preparation method for B8631 and its solubility?

    1 answer
    1. The product B8631 should be considered a crude extract. In terms of solubility, quality control testing indicates that this product is tested for solubility in 80% ethanol at a concentration of 10 mg/mL, resulting in a slightly hazy to very hazy, very dark yellow solution. The color of the solution is within the acceptable range specified in the current Specification Sheet.

      Helpful?

  3. I would like to know if this product contains taurocholic acid and sodium taurodeoxycholate and how much of each. Thank you

    1 answer
    1. Taurocholic acid and sodium taurodeoxycholate are not tested on a lot to lot basis for this product. However, historical data reveals that typically this material contains approximately 4-7% taurodeoxycholic acid and 30-40% taurocholic acid.

      Helpful?

  4. What are the best solvents that can be used to solubilize this product?

    1 answer
    1. This product is soluble in 80% ethanol at 10mg/mL which yields a slightly hazy to very hazy solution. This material may also be dissolved in water at 50mg/mL. However, this will yield a turbid solution.

      Helpful?

  5. What is the content of cholesterol (it should mostly be free cholesterol) in B8381 and B8631?

    1 answer
    1. Neither B8381 nor B8631 have been evaluated for cholesterol content. The approximate amount of known components of both are listed below:
      8.0 - 9.5 % cholic acid
      0.9 - 1.1 % deoxycholic acid
      32 - 36 % conjugated cholic acid
      11 - 13 % conjugated deoxycholic acid

      Helpful?

  6. What is the proper temperature for storage?

    1 answer
    1. This product is stored at room temperature.

      Helpful?

Reviews

No rating value

Active Filters