Skip to Content
Merck
All Photos(1)

Documents

N9535

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide, reduced disodium salt

~98%, pkg of 25-30 mg (per vial)

Synonym(s):

β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2
CAS Number:
Molecular Weight:
709.40
Beilstein:
5230241
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~98%

Quality Level

form

powder

packaging

pkg of 25-30 mg (per vial)

SMILES string

[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O

InChI

1S/C21H29N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;/q;2*+1/p-2/t10-,11+,13-,14+,15-,16+,20-,21+;;/m0../s1

InChI key

QRGNQKGQENGQSE-QUWMEQBESA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).

Biochem/physiol Actions

Electron donor

Reconstitution

Solutions should be prepared fresh and used promptly.

Other Notes

Packaged based on NADH content as determined by UV-Vis.
This is the common form of NADH; do not confuse with α-NADH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weihai Ying
Frontiers in bioscience : a journal and virtual library, 11, 3129-3148 (2006-05-25)
Increasing evidence has indicated that NAD+ and NADH play critical roles not only in energy metabolism, but also in cell death and various cellular functions including regulation of calcium homeostasis and gene expression. It has also been indicated that NAD+
Shin-Ichiro Imai
Biochimica et biophysica acta, 1804(8), 1584-1590 (2009-11-10)
SIR2 (silent information regulator 2) proteins, now called "sirtuins," are an evolutionarily conserved family of NAD-dependent protein deacetylases/ADP-ribosyltransferases. Sirtuins have recently attracted major attention in the field of aging research, and it has been demonstrated that SIR2 and its orthologs
Richard P Ebstein et al.
FEBS letters, 585(11), 1529-1536 (2011-05-12)
Increasing evidence suggests that the nonapeptide, oxytocin (OT), helps shape social and affiliative behaviors not only in lower mammals but also in humans. Recently, an essential mediator of brain OT release has been discovered, ADP-ribosyl cyclase and/or CD38. We have
Xiaoling Li et al.
International journal of biological sciences, 7(5), 575-587 (2011-05-27)
Sirtuins are highly conserved NAD+-dependent protein deacetylases and/or ADP-ribosyltransferases that can extend the lifespan of several lower model organisms including yeast, worms and flies. The seven mammalian sirtuins, SIRT1 to SIRT7, have emerged as key metabolic sensors that directly link
Hening Lin
Organic & biomolecular chemistry, 5(16), 2541-2554 (2007-11-21)
ADP-ribosylation using nicotinamide adenine dinucleotide (NAD+) is an important type of enzymatic reaction that affects many biological processes. A brief introductory review is given here to various ADP-ribosyltransferases, including poly(ADP-ribose) polymerase (PARPs), mono(ADP-ribosyl)-transferases (ARTs), NAD(+)-dependent deacetylases (sirtuins), tRNA 2'-phosphotransferases, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service