Skip to Content
Merck
All Photos(4)

Key Documents

N5755

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate hydrate

Synonym(s):

β-NADP, Coenzyme II, NADP, TPN, Triphosphopyridine nucleotide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28N7O17P3 · xH2O
CAS Number:
Molecular Weight:
743.41 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥95% (HPLC)
≥95% (spectrophotometric assay)

Quality Level

form

powder

solubility

H2O: soluble 50 mg/mL, clear, colorless to faintly yellow

cation traces

sodium and potassium: ≤0.04%

storage temp.

−20°C

SMILES string

O.NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]3O[C@H]([C@H](OP(O)(O)=O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O17P3.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);1H2/t10-,11-,13-,14-,15-,16-,20-,21-;/m1./s1

InChI key

ZKJOXOJMGXFSPF-QYZPTAICSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide phosphate hydrate is suitable for use in:
  • the measurement of Glucose-6-phosphate dehydrogenase activity
  • the Cytochrome P450 3A4 assay as a part of NADPH-regenerating system
  • the Cytochrome P450 2D6 assay as a part of NADPH-regenerating system
  • the determination of Glucose-6-phosphate content

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway (PPP).
Electron acceptor

Other Notes

Packaged based on solid weight.
This is a common form of NADP.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M Lilley et al.
Oecologia, 175(3), 811-823 (2014-05-20)
Harmful reactive oxygen species (ROS) produced during metabolism and immune responses are neutralized in part by a powerful enzymatic antioxidant system. Inter-species variability in the baseline activity of antioxidant enzymes may be explained by a variety of life history traits.
Raoul G Rosenthal et al.
Nature chemical biology, 10(1), 50-55 (2013-11-19)
The pyridine nucleotides NADH and NADPH (NAD(P)H) are ubiquitous redox coenzymes that are present in all living cells. Although about 16% of all characterized enzymes use pyridine nucleotides as hydride donors or acceptors during catalysis, a detailed understanding of how
Bent H Hellum et al.
Basic & clinical pharmacology & toxicology, 102(5), 466-475 (2008-03-12)
Extracts of six commonly used commercially available herbal products, St. John's wort, common valerian, common sage, Ginkgo biloba, Echinacea purpurea and horse chestnut were investigated for their in vitro inhibitory potential of CYP3A4 mediated metabolism and P-glycoprotein efflux transport activity.
William B. Wood
Biochemistry: A Problems Approach, 195-195 null
Berg, J.M., et al.
Biochemistry (5th Edition) (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service