Skip to Content
Merck

N8129

β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate

≥97% (HPLC)

Synonym(s):

β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2 · xH2O
CAS Number:
Molecular Weight:
709.40 (anhydrous basis)
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
210-123-3
MDL number:
Form:
powder
Assay:
≥97% (HPLC), ≥97% (spectrophotometric assay)
Solubility:
water: 712 g/L at 22.5 °C, 10 mM NaOH: soluble-100 mg/mL, clear to slightly hazy, faintly yellow to greenish-yellow
Biological source:
yeast
Color:
white to light yellow
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

yeast

Quality Segment

assay

≥97% (HPLC), ≥97% (spectrophotometric assay)

form

powder

mol wt

709.40 g/mol

color

white to light yellow

solubility

water: 712 g/L at 22.5 °C, 10 mM NaOH: soluble-100 mg/mL, clear to slightly hazy, faintly yellow to greenish-yellow

density

1.995 g/cm3 at 20 °C

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H29N7O14P2.2Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;;1H2/q;2*+1;/p-2/t10-,11-,13-,14-,15-,16-,20-,21-;;;/m1.../s1

InChI key

FWBNCIFELNNJCX-UHOVGGJYSA-L

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).
As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.

Biochem/physiol Actions

NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, β-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). It participates in cell signaling events as well, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response. The NAD+/NADH dependent sirtuins play key roles in stress responses during events involving energy metabolism, with implications in cancer biology, diabetes and neurodegenerative disease.
As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.

Packaging

Packaged by solid weight.


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
N8129-1G04061835573646
N8129-5G04061835516193
N8129-50MG04061835506521
N8129-100MG04061835573639
N8129-500MG04061835573653
N8129-5MG04061833492987