Skip to Content
Merck
All Photos(1)

Key Documents

N7785

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt

vial of 10 mg, ~95%

Synonym(s):

β-NADPH, 2′-NADPH, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N7Na4O17P3
Molecular Weight:
833.35
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~95%

Quality Level

form

powder

packaging

vial of 10 mg

SMILES string

[Na+].[Na+].[Na+].[Na+].[P](=O)([O-])([O-])O[C@H]1[C@@H](O[C@@H]([C@H]1O)CO[P](=O)([O-])O[P](=O)([O-])OC[C@@H]4O[C@@H]([C@H]([C@H]4O)O)N5C=C(CC=C5)C(=O)N)[n]2c3ncnc(c3nc2)N

InChI

1S/C21H30N7O17P3.4Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;;;/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);;;;/q;4*+1/p-4/t10-,11+,13-,14+,15-,16+,20-,21+;;;;/m0..../s1

InChI key

WYWWVJHQDVCHKF-MPUNMZHWSA-J

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway (PPP).
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt has been used in the Griess reagent assay of culture medium to reduce nitrate to nitrite and in glutathione peroxidase assay.

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivo by the pentose phosphate pathway (PPP).
Electron donor; cofactor for many redox enzymes including nitric oxide synthetase

Preparation Note

Chemically reduced

Other Notes

Packaged based on NADPH content as determined by UV-Vis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium potentiate oxidative burden and reduced antioxidant status in different rat organs system
Joshi DK, et al.
Int J Toxicol Pharmacol Res, 5(1), 9-14 (2013)
Assays for nitric oxide expression
Stone WL, et al.
Methods in Molecular Biology, 245-256 (2006)
Cristina García-Díaz et al.
Frontiers in neuroanatomy, 15, 674649-674649 (2021-07-10)
Spatial learning and memory processes depend on anatomical and functional interactions between the hippocampus and the entorhinal cortex. A key neurophysiological component of these processes is hippocampal theta rhythm, which can be driven from subcortical areas including the pontine nucleus
Xiao-Chuan Cai et al.
eLife, 8 (2019-10-29)
CARM1 is a cancer-relevant protein arginine methyltransferase that regulates many aspects of transcription. Its pharmacological inhibition is a promising anti-cancer strategy. Here SKI-73 (6a in this work) is presented as a CARM1 chemical probe with pro-drug properties. SKI-73 (6a) can
Grant R Drummond et al.
Nature reviews. Drug discovery, 10(6), 453-471 (2011-06-02)
NADPH oxidases are a family of enzymes that generate reactive oxygen species (ROS). The NOX1 (NADPH oxidase 1) and NOX2 oxidases are the major sources of ROS in the artery wall in conditions such as hypertension, hypercholesterolaemia, diabetes and ageing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service