Skip to Content
Merck
All Photos(2)

Key Documents

N5655

Sigma-Aldrich

Nicotinic acid adenine dinucleotide phosphate sodium salt

≥92.5%

Synonym(s):

β-NAADP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N6O18P3 · xNa+
CAS Number:
Molecular Weight:
744.39 (free acid basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥92.5%

form

solid

storage temp.

−20°C

SMILES string

[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OCC4O[C@@H]([C@@H](O)[C@H]4O)[n+]5cccc(c5)C(O)=O)[C@@H](O)[C@H]3OP(O)(O)=O

InChI

1S/C21H27N6O18P3.Na/c22-17-12-18(24-7-23-17)27(8-25-12)20-16(44-46(33,34)35)14(29)11(43-20)6-41-48(38,39)45-47(36,37)40-5-10-13(28)15(30)19(42-10)26-3-1-2-9(4-26)21(31)32;/h1-4,7-8,10-11,13-16,19-20,28-30H,5-6H2,(H6-,22,23,24,31,32,33,34,35,36,37,38,39);/q;+1/p-1/t10?,11-,13+,14-,15+,16-,19+,20-;/m1./s1

InChI key

KKCRODBDXQTAPI-NZNNTXIPSA-M

General description

NAADP induces Ca2+ release from the eggs of sea urchin, when injected into the cell. It can be enzymatically produced from nicotinamide adenine dinucleotide phosphate (β-NADP) with the help of ADP-ribosyl cyclase.

Application

Nicotinic acid adenine dinucleotide phosphate sodium salt has been used for agonist injection into unfertilized eggs and embryos.

Biochem/physiol Actions

Potent activator of intracellular Ca2+ release via an IP3 and cyclic ADP ribose-independent mechanism.

Other Notes

Note: Should not be confused with nicotinamide adenine dinucleotide phosphate (NADP).

Preparation Note

Enzymatically prepared analog of β-NADP+.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E N Chini et al.
The Journal of biological chemistry, 270(7), 3216-3223 (1995-02-17)
Transient fluxes of intracellular ionized calcium (Ca2+) from intracellular stores are integral components of regulatory signaling pathways operating in numerous biological regulations, including in early stages of egg fertilization. Therefore, we explored whether NADP, which is rapidly generated by phosphorylation
Patricia Gómez-Suaga et al.
Autophagy, 8(4), 692-693 (2012-03-24)
Alterations in autophagy are thought to underlie various neurodegenerative diseases including Parkinson disease (PD). Previous studies have indicated that the PD gene leucine rich repeat kinase 2 (LRRK2) is involved in this process, but its mechanism of action has remained
H C Lee et al.
The Journal of biological chemistry, 270(5), 2152-2157 (1995-02-03)
We have previously shown that alkaline treatment of NADP generates a derivative which can mobilize Ca2+ from sea urchin egg homogenates (Clapper, D. L., Walseth, T. F., Dargie, P. J., and Lee, H. C. (1987) J. Biol. Chem. 262, 9561-9568).
Slow calcium waves mediate furrow microtubule reorganization and germ plasm compaction in the early zebrafish embryo
Eno C, et al.
Development (2018)
Parvinder K Aley et al.
The Journal of biological chemistry, 288(16), 10986-10993 (2013-03-08)
Nicotinic acid adenine dinucleotide phosphate (NAADP) is increasingly being demonstrated to be involved in calcium signaling in many cell types and species. Although it has been shown to play a role in smooth muscle cell contraction in several tissues, nothing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service