Skip to Content
Merck
All Photos(3)

Documents

M2514

Sigma-Aldrich

4-Methylumbelliferyl heptanoate

≥95% (GC)

Synonym(s):

4-Methylumbelliferyl enanthate, Heptanoic acid 4-methylumbelliferyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O4
CAS Number:
Molecular Weight:
288.34
Beilstein:
1256801
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (GC)

form

powder

mp

41-42 °C (lit.)

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 312 nm in methanol
λex 360 nm; λem 449 nm (Reaction product)

storage temp.

−20°C

SMILES string

CCCCCCC(=O)Oc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C17H20O4/c1-3-4-5-6-7-16(18)20-13-8-9-14-12(2)10-17(19)21-15(14)11-13/h8-11H,3-7H2,1-2H3

InChI key

FFNBFZWIBOIPIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Fiksdal et al.
Applied and environmental microbiology, 55(9), 2424-2427 (1989-09-01)
The production of an enzyme, 4-methylumbelliferyl heptanoate hydrolase, in Escherichia coli exposed to enriched and nonenriched seawater was studied. In all media, except for seawater with no or very small amounts of organic material and seawater enriched with peptone, 4-methylumbelliferyl
S Krüger-Krasagakes et al.
Journal of immunological methods, 156(1), 1-8 (1992-11-25)
In order to measure cell mediated cytotoxicity to adherent growing cell lines in vitro more rapidly and conveniently, a fluorometric microassay was developed and results were compared with those obtained by the 51Cr release assay. The fluorometric method is based
L Virág et al.
Journal of immunological methods, 185(2), 199-208 (1995-09-25)
A fluorimetric method using 4-methylumbelliferyl heptanoate (MUH) has been developed for detecting cell-mediated cytotoxicity and cell proliferation. The assay is based on the hydrolysis of the fluorochrome (MUH) by intracellular esterases of viable cells resulting in the production of highly
E N Dotsika et al.
Journal of immunological methods, 105(1), 55-62 (1987-12-04)
A microplate method for assessing cell growth and viability based on the hydrolysis of fluorogenic substrates by cell esterases has been investigated. Living cells incubated with fluorescein diacetate or 4-methylumbelliferyl heptanoate generate a fluorescent product which is proportional to the
C C Zouboulis et al.
Melanoma research, 1(2), 91-95 (1991-06-01)
A simple, rapid and reproducible assay for the determination of melanoma cell proliferation in vitro is described, based on the hydrolysis of a fluorogenic substrate by cell esterases in the cytoplasm of living cells. Human melanoma cells were cultured at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service