Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-619-9
MDL number:
Specific activity:
≥20,000 units/mg protein
Pricing and availability is not currently available.
type
Type VI-S
Quality Segment
form
lyophilized powder
specific activity
≥20,000 units/mg protein
composition
Protein, 40-70%
shipped in
wet ice
storage temp.
−20°C
InChI
1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;
InChI key
QWZUIMCIEOCSJF-CHHCPSLASA-N
General description
Lipase from porcine pancreas (PPL) or porcine pancreatic lipase is a small globular protein with N-terminal α/β type fold. It comprises catalytic triad Ser, Asp, and His residues and a C-terminal β-sandwich domain. PPL belongs to a subclass of the carboxylesterases family.[1]
Application
Lipase from porcine pancreas has been used:
- in lipase inhibition assay with plant extracts[2]
- in in vitro digestion gelatinized starch/fiber mixtures[3]
- with simulated intestinal fluid to mimic the digestive tract condition of by L. fermentum PC1 in fermented oats[4]
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Biochem/physiol Actions
Lipase from porcine pancreas (PPL) catalyzes the hydrolysis of fats (lipids). It plays an essential role in the digestion, transport, and processing of dietary lipids (e.g. triglycerides, fats, oils) in most living organisms.[1] PPL is cost-effective and is useful in biotransformation reactions. It displays high selectivity, and catalytic activity. PPL is also used in the detergent formulation for removing oil stains from clothes.
Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions. They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
Analysis Note
Protein determined by biuret.
Other Notes
One unit will hydrolyze 1.0 microequivalent of fatty acid from a triglyceride in 1 hr at pH 7.7 at 37 °C using olive oil.
1 of 1
This Item | |||
|---|---|---|---|
| specific activity ≥20,000 units/mg protein | specific activity ≥40,000 units/mg protein | specific activity 15-25 U/mg | specific activity ≥2 U/mg |
| form lyophilized powder | form lyophilized powder | form lyophilized solid, powder (fine) | form powder |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| Quality Level 300 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| type Type VI-S | type - | type - | type - |
Still not finding the right product?
Explore all of our products under Lipase from porcine pancreas
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




