Skip to Content
Merck
4-Methylumbelliferyl heptanoate ≥95% (GC)

4-Methylumbelliferyl heptanoate

4-Methylumbelliferyl heptanoate ≥95% (GC)
Synonym(s):
4-Methylumbelliferyl enanthate, Heptanoic acid 4-methylumbelliferyl ester
Empirical Formula (Hill Notation):
C17H20O4
CAS Number:
Molecular Weight:
288.34
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing