Skip to Content
Merck
All Photos(1)

Key Documents

M0883

Sigma-Aldrich

4-Methylumbelliferyl acetate

esterase substrate

Synonym(s):

MU-Ac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O4
CAS Number:
Molecular Weight:
218.21
Beilstein:
189667
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (TLC)

form

powder

mp

149-150 °C (lit.)

solubility

chloroform: 100 mg/mL, clear, colorless

fluorescence

λex 312 nm in methanol
λex 360 nm; λem 499 nm (Reaction product)

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C12H10O4/c1-7-5-12(14)16-11-6-9(15-8(2)13)3-4-10(7)11/h3-6H,1-2H3

InChI key

HXVZGASCDAGAPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Frimer et al.
Free radical biology & medicine, 20(6), 843-852 (1996-01-01)
--2-(Dimethylamino) fluorene (1a) and 5-benzoyloxy-2,3,7,8,12,13,17,18-octaethylporphyrin (4) react with superoxide anion radical (generated from KO2/18-crown-6 polyether) in aprotic media. Yet, when incorporated into the lipid bilayer of dimyristoyl phosphatidylcholine liposomes, these two substrates are inert to superoxide, generated enzymatically (xanthine oxidase/acetaldehyde)
Enzyme kinetic studies in cell populations using fluorogenic substrates and flow cytometric techniques.
J V Watson
Cytometry, 1(2), 143-151 (1980-09-01)
H G Raj et al.
Teratogenesis, carcinogenesis, and mutagenesis, 21(2), 181-187 (2001-02-27)
Our earlier studies documented the ability of 7,8-diacetoxy-4-methylcoumarin (DAMC) to cause irreversible inhibition of cytochrome P-450 linked mixed function oxidases (MFO) mediated by membrane bound DAMC: protein transacetylase. Since P-450 catalyzed oxidation of benzene is crucial to its toxic effects
H A Berman et al.
Biochemistry, 29(47), 10640-10649 (1990-11-27)
This paper examines covalent reactivity of AchE with respect to cationic and uncharged methylphosphonates and substrates in the absence and presence of cationic ligands selective for the active center and the peripheral anionic site. The organophosphorus inhibitors are enantiomeric alkyl
Abha Kathuria et al.
Bioorganic & medicinal chemistry, 17(4), 1550-1556 (2009-02-03)
Calreticulin Transacetylase (CRTAase) catalyzes the transfer of acetyl groups from polyphenolic acetates (PAs) to the receptor proteins and modulates their biological activities. CRTAase was conveniently assayed by the irreversible inhibition of cytosolic glutathione S-transferase (GST) by the model acetoxycoumarin, 7,8-diacetoxy-4-methylcoumarin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service