Skip to Content
Merck
All Photos(1)

Key Documents

M2319

Sigma-Aldrich

Mefloquine hydrochloride

≥98% (HPLC), powder

Synonym(s):

(AS)-rel-a-(2R)-2-Piperidinyl-2,8-bis(trifluoromethyl)-4-quinolinemethanol monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClF6N2O
CAS Number:
Molecular Weight:
414.77
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

DMSO: soluble 38 mg/mL
H2O: insoluble

SMILES string

Cl[H].[H][C@@]1(CCCCN1)[C@@H](O)c2cc(nc3c(cccc23)C(F)(F)F)C(F)(F)F

InChI

1S/C17H16F6N2O.ClH/c18-16(19,20)11-5-3-4-9-10(15(26)12-6-1-2-7-24-12)8-13(17(21,22)23)25-14(9)11;/h3-5,8,12,15,24,26H,1-2,6-7H2;1H/t12-,15+;/m1./s1

InChI key

WESWYMRNZNDGBX-YLCXCWDSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mefloquine hydrochloride has been used:
  • to treat cochlear organotypic cultures with various doses to evaluate its role in cellular pathway involved in apoptosis
  • in screening for in vitro antischistosomal activity
  • in cytotoxicity assay of glioblastoma cells

Biochem/physiol Actions

Mefloquine is broadly used as an antimalarial drug. It inhibits 80S ribosomes of Plasmodium. It has numerous side effects like anxiety, dizziness, tremor, headache, and hearing loss. Mefloquine damages cochlear and vestibular hair cells through apoptosis. It also damages spiral ganglion neurons, degenerates cortical neuron and disrupts neuronal calcium homeostasis.
Blocker of gap junction channels Cx36 and Cx50.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Zaloumis et al.
Malaria journal, 11, 303-303 (2012-08-31)
Mechanistic within-host models relating blood anti-malarial drug concentrations with the parasite-time profile help in assessing dosing schedules and partner drugs for new anti-malarial treatments. A comprehensive simulation study to assess the utility of a stage-specific pharmacokinetic-pharmacodynamic (PK-PD) model for predicting
Regulation of hypoxia-induced autophagy in glioblastoma involves ATG9A
Rahim SAA, et al.
British Journal of Cancer, 117(6), 813-813 (2017)
Chanaki Amaratunga et al.
The Lancet. Infectious diseases, 12(11), 851-858 (2012-09-04)
Artemisinin-resistant Plasmodium falciparum has been reported in Pailin, western Cambodia, detected as a slow parasite clearance rate in vivo. Emergence of this phenotype in western Thailand and possibly elsewhere threatens to compromise the effectiveness of all artemisinin-based combination therapies. Parasite
Ryutaro Tanizaki et al.
Malaria journal, 12, 128-128 (2013-04-17)
This is the first case of Plasmodium knowlesi infection in a Japanese traveller returning from Malaysia. In September 2012, a previously healthy 35-year-old Japanese man presented to National Center for Global Health and Medicine in Tokyo with a two-day history
Addition of exogenous NAD+ prevents mefloquine-induced neuroaxonal and hair cell degeneration through reduction of caspase-3-mediated apoptosis in cochlear organotypic cultures
Ding D, et al.
Testing, 8(11), e79817-e79817 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service