Skip to Content
Merck
All Photos(1)

Key Documents

PHR1705

Supelco

Mefloquine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Mefloquine hydrochloride, MQ, (AS)-rel-a-(2R)-2-Piperidinyl-2,8-bis(trifluoromethyl)-4-quinolinemethanol monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClF6N2O
CAS Number:
Molecular Weight:
414.77
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M0253000
traceable to USP 1379059

API family

mefloquine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].[H][C@@]1(CCCCN1)[C@@H](O)c2cc(nc3c(cccc23)C(F)(F)F)C(F)(F)F

InChI

1S/C17H16F6N2O.ClH/c18-16(19,20)11-5-3-4-9-10(15(26)12-6-1-2-7-24-12)8-13(17(21,22)23)25-14(9)11;/h3-5,8,12,15,24,26H,1-2,6-7H2;1H/t12-,15+;/m1./s1

InChI key

WESWYMRNZNDGBX-YLCXCWDSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

Mefloquine Hydrochloride may be used as a pharmaceutical reference standard for the determination of mefloquine in pharmaceutical formulations and plasma samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Blocker of gap junction channels Cx36 and Cx50.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3945 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a generic micellar electrokinetic chromatography method for the separation of 15 antimalarial drugs as a tool to detect medicine counterfeiting.
Lamalle C, et al.
Electrophoresis, 33(11), 1669-1678 (2012)
Development and validation of a reversed-phase LC method for analysing potentially counterfeit antimalarial medicines.
Gaudiano MC, et al.
Journal of Pharmaceutical and Biomedical Analysis, 42(1), 132-135 (2006)
Screening study of SFC critical method parameters for the determination of pharmaceutical compounds.
Dispas A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 125(31), 339-354 (2016)
Innovative high-performance liquid chromatography method development for the screening of 19 antimalarial drugs based on a generic approach, using design of experiments, independent component analysis and design space.
Debrus B, et al.
Journal of Chromatography A, 1218(31), 5205-5215 (2011)
An analytical method with a single extraction procedure and two separate high performance liquid chromatographic systems for the determination of artesunate, dihydroartemisinin and mefloquine in human plasma for application in clinical pharmacological studies of the drug combination.
Lai CS, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 857(2), 308-314 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service