Skip to Content
Merck
All Photos(1)

Documents

M103

Sigma-Aldrich

1-Methyl-4-(2′-methylphenyl)-1,2,3,6-tetrahydropyridine hydrochloride

solid

Synonym(s):

2′-Methyl-MPTP hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N · HCl
CAS Number:
Molecular Weight:
223.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

H2O: soluble
ethanol: soluble

SMILES string

Cl[H].CN1CCC(=CC1)c2ccccc2C

InChI

1S/C13H17N.ClH/c1-11-5-3-4-6-13(11)12-7-9-14(2)10-8-12;/h3-7H,8-10H2,1-2H3;1H

InChI key

QLIQCKBPYIWPTE-UHFFFAOYSA-N

Application

1-Methyl-4-(2′-methylphenyl)-1,2,3,6-tetrahydropyridine hydrochloride has been used to study its effect on core temperature in C57BL/6J mice.

Biochem/physiol Actions

1-Methyl-4-(2′-methylphenyl)-1,2,3,6-tetrahydropyridine hydrochloride (MPTP) is a tertiary amine. It is widely used to induce experimental parkinsonism in mice.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T May
Neuroscience letters, 162(1-2), 55-58 (1993-11-12)
The in vitro binding of [3H]pargyline and [3H]harman ([3H]1-methyl-beta-carboline) to monoamine oxidase A (MAO-A; EC 1.4.3.4) on membranes of rat cerebral cortex was evaluated. Displacement of the [3H]pargyline binding on MAO-A (L(-)-deprenyl suppressed binding to MAO-B) by harman, 1-methyl-4-phenylpyridinium (MPP+)
P K Sonsalla et al.
The Journal of pharmacology and experimental therapeutics, 242(3), 850-857 (1987-09-01)
1-Methyl-4-(2'-methylphenyl)-1,2,3,6-tetrahydropyridine (2'CH3-MPTP) was shown previously to be a more potent neurotoxicant than 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) in mice. The present investigation was conducted to determine possible reasons for the greater potency of 2'CH3-MPTP and to determine if its neurotoxic action might be
N Yoshimura et al.
Neuropharmacology, 32(4), 315-321 (1993-04-01)
A pharmacological study using monkeys, in which parkinsonism was induced by 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP), was undertaken to elucidate the mechanism underlying urinary bladder dysfunctions in Parkinson's disease. Under ketamine anesthesia, cystometrograms showed that, in MPTP-treated monkeys, a contraction of the urinary
F F Cruz-Sanchez et al.
Molecular and chemical neuropathology, 19(1-2), 163-176 (1993-05-01)
In order to compare the recovery capacity of the nigrostriatal system between adult and old mice, MPTP hydrochloride was administered to 48 BL/C57 male mice, which were sacrificed 24 h or 10 d after the second dose. The animals were
Voloshin MYa et al.
Neuroscience, 53(3), 845-854 (1993-04-01)
Activity of entopeduncular neurons was studied in chronic experiments on cats during performance of instrumental movement: pedal pressing and holding. One-hundred and twenty-four neurons were extracellularly investigated in intact animals and 81 neurons in cats treated with N-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) (5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service