Skip to Content
Merck
All Photos(5)

Key Documents

H4381

Sigma-Aldrich

6-Hydroxydopamine hydrochloride

≥97% (titration), powder, neurotoxin

Synonym(s):

2,4,5-Trihydroxyphenethylamine hydrochloride, 2,5-Dihydroxytyramine hydrochloride, 2-(2,4,5-Trihydroxyphenyl)ethylamine hydrochloride, 6-OHDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
4274007
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Product Name

6-Hydroxydopamine hydrochloride, ≥97% (titration), powder

Assay

≥97% (titration)

form

powder

color

off-white to brown

mp

232-233 °C (dec.) (lit.)

solubility

H2O: >50 mg/mL, clear, yellow to brown

storage temp.

room temp

SMILES string

Cl.NCCc1cc(O)c(O)cc1O

InChI

1S/C8H11NO3.ClH/c9-2-1-5-3-7(11)8(12)4-6(5)10;/h3-4,10-12H,1-2,9H2;1H

InChI key

QLMRJHFAGVFUAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Hydroxydopamine hydrochloride (6-OHDA) is a neurotoxin, commonly used to induce Parkinson′s disease (PD). 6-OHDA causes death of dopaminergic neurons in substantia nigra pars compacta progressively mimicking PD. 6-OHDA is highly oxidizable and cannot cross blood brain barrier. 6-OHDA exerts cytotoxicity by generating reactive oxygen species, initiating cellular stress and cell death. 6-OHDA leads to neuronal cell death in many in vitro models like primary neuronal culture, human neuroblastoma cell line and rat adrenal pheochromocytoma cell line, PC12.

Application

6-hydroxydopamine hydrochloride has been used:
  • to induce Parkinson′s disease (PD) in rats
  • to analyse cytotoxic effect of 6-OHDA on PC12 cell line
  • to induce noradrenergic (NA) neuron deletion from the locus-coeruleus

Biochem/physiol Actions

Neurotoxin that destroys catecholaminergic terminals.

Caution

Hygroscopic

Reconstitution

Dissolve in oxygen-free water containing 0.1% sodium metabisulfite or other antioxidants.
Solutions should be freshly prepared and protected from exposure to light. Solutions turn red as it oxidizes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kryspin Andrzejewski et al.
Journal of biomedical science, 24(1), 24-24 (2017-03-30)
Malfunctioning of the serotonergic system in Parkinson's disease may contribute to non-motor symptoms such as respiratory complications. Thus the aim of our study was to investigate the role of serotonin 5-HT Wistar rats were lesioned unilaterally with double 6-hydroxydopamine (6-OHDA)
Molecular mechanisms of 6-hydroxydopamine-induced cytotoxicity in PC12 cells: involvement of hydrogen peroxide-dependent and-independent action
Saito Y, et al.
Free Radical Biology & Medicine, 42(5), 675-685 (2007)
Human induced pluripotent stem cell-derived neurons improve motor asymmetry in a 6-hydroxydopamine-induced rat model of Parkinson's disease
Han F, et al.
Cytotherapy, 17(5), 665-679 (2015)
The 6-hydroxydopamine model and parkinsonian pathophysiology: Novel findings in an older model
Hernandez-Baltazar D, et al.
Neurologia (Barcelona, Spain), 32(8), 533-539 (2017)
Pingping Song et al.
Protein & cell, 7(2), 114-129 (2016-01-10)
Mutations or inactivation of parkin, an E3 ubiquitin ligase, are associated with familial form or sporadic Parkinson's disease (PD), respectively, which manifested with the selective vulnerability of neuronal cells in substantia nigra (SN) and striatum (STR) regions. However, the underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service