Skip to Content
Merck
All Photos(1)

Documents

SMB00173

Sigma-Aldrich

Prunasin

≥90% (LC/MS-ELSD)

Synonym(s):

D-Mandelonitrile β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO6
CAS Number:
Molecular Weight:
295.29
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OCC1OC(OC(C#N)c2ccccc2)C(O)C(O)C1O

InChI

1S/C14H17NO6/c15-6-9(8-4-2-1-3-5-8)20-14-13(19)12(18)11(17)10(7-16)21-14/h1-5,9-14,16-19H,7H2

InChI key

ZKSZEJFBGODIJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Yamaguchi et al.
Bioscience, biotechnology, and biochemistry, 82(11), 2021-2029 (2018-07-22)
Japanese apricot, Prunus mume Sieb. et Zucc., biosynthesizes the l-phenylalanine-derived cyanogenic glucosides prunasin and amygdalin. Prunasin has biological properties such as anti-inflammation, but plant extraction and chemical synthesis are impractical. In this study, we identified and characterized UGT85A47 from Japanese
Tan Hooi Poay et al.
Phytochemical analysis : PCA, 22(6), 516-525 (2011-04-16)
Phyllagathis rotundifolia (Jack) Bl. (Melastomataceae) is a creeping herb found in Peninsular Malaysia and Sumatra. Traditionally, a decoction of the leaves is used in the treatment of malaria, fever and stomach ache. To provide ESI-MS(n) data which are applicable for
Raquel Sánchez-Pérez et al.
Plant physiology, 146(3), 1040-1052 (2008-01-15)
Bitterness in almond (Prunus dulcis) is determined by the content of the cyanogenic diglucoside amygdalin. The ability to synthesize and degrade prunasin and amygdalin in the almond kernel was studied throughout the growth season using four different genotypes for bitterness.
Jason Q D Goodger et al.
Oecologia, 153(4), 799-808 (2007-07-03)
Many studies have shown that similarly aged plants within a species or population can vary markedly in the concentration of defence compounds they deploy to protect themselves from herbivores. Some studies have also shown that the concentration of these compounds
Rebecca E Miller et al.
Phytochemistry, 67(1), 43-51 (2005-11-26)
The cyanogenic diglycoside lucumin ((R)-mandelonitrile-beta-D-primeveroside) and monoglucoside prunasin ((R)-mandelonitrile-beta-D-glucoside) were isolated from the foliage of the rare Australian rainforest tree species Clerodendrum grayi (Lamiaceae). This is the first reported isolation of the diglycoside lucumin from vegetative tissue (foliage), and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service