Skip to Content
Merck
All Photos(4)

Key Documents

M0387

Sigma-Aldrich

Methyl cellulose

viscosity: 1,500 cP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

wood (pulp cellulose)

form

powder

color

white to off-white

viscosity

1,200-1,800 cP, 2 % in water(20 °C)

storage temp.

room temp

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl cellulose is a tough, non-toxic material. It is odorless and tasteless. Hence, methyl cellulose is widely used as an adhesive in the food packaging industry. In water-soluble adhesives, it is utilized as a thickening agent. Methyl cellulose is also used as a viscosity control agent. In the cosmetic industry, it is used as an emulsifier, binder and stabilizer.

Application

Methyl cellulose has been used:
  • as a component of an embryo medium to culture zebrafish embryos for motility assays
  • to mount haploid/tetraploid zebrafish embryos on slides for live imaging
  • to enhance the viscosity of artificial seawater to study the flagellar waves of Ciona spermatozoa

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The zebrafish mutant lessen: an experimental model for congenital enteric neuropathies
Uyttebroek L, et al.
Neurogastroenterology and Motility, 28(3), 345-357 (2016)
Ivana Hermanova et al.
The Journal of experimental medicine, 217(6) (2020-03-29)
Gene dosage is a key defining factor to understand cancer pathogenesis and progression, which requires the development of experimental models that aid better deconstruction of the disease. Here, we model an aggressive form of prostate cancer and show the unconventional
Mechanical constraint converts planar waves into helices on tunicate and sea urchin sperm flagella
Ishijima S
Cell Structure and Function, 1-23 (2011)
Handbook of Adhesives and Surface Preparation (2011)
Chia-Hao Hsu et al.
Scientific reports, 5, 10673-10673 (2015-06-04)
mib(nn2002), found from an allele screen, showed early segmentation defect and severe cell death phenotypes, which are different from previously known mib mutants. Despite distinct morphological phenotypes, the typical mib molecular phenotypes: her4 down-regulation, neurogenic phenotype and cold sensitive dlc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service