Skip to Content
Merck

Skip To

M6385

Methyl cellulose

viscosity: 25 cP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

color

white to off-white

viscosity

20-30 cP, 2 % in water(20 °C)

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

General description

Methyl cellulose (MC) is a simple derivative of cellulose.

Application

Methyl cellulose has been used:
  • as a supplement for culturing multicellular spheroids of SH-SY5Y cells[1]
  • as a supplement for culturing tumor cell spheroids[2]
  • to generate and study the rheological behavior of aqueous mullite-albumin-methylcellulose systems

Biochem/physiol Actions

Methyl cellulose (MC) acts as a laxative and is hydrophilic. It has therapeutic effects against constipation, irritable bowel syndrome, and diverticulosis due to its bulk-forming and stool softening properties. MC has the potential to treat patients with ileostomies and colostomies. It may also be used as a stabilizer and emulsifier. MC is a component of eye drops and contact lens solutions.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
94378M0430M7027
description

viscosity: 25 cP

description

viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)

description

meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP

description

viscosity: 15 cP, BioReagent, suitable for cell culture

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

color

white to off-white

color

-

color

-

color

-

viscosity

20-30 cP, 2 % in water(20 °C)

viscosity

3000-5500 mPa.s, 2 % in H2O(20 °C)

viscosity

320-480 cP, 2 % in water(20 °C)

viscosity

12-18 cP, 2 % in H2O(20 °C)


Still not finding the right product?

Explore all of our products under Methyl cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Can I know the Mw range?

    1 answer
    1. The average molecular weight is 17000 Daltons.

      Helpful?

  2. What is the degree of substitution of this material?

    1 answer
    1. The degree of substitution for this product ranges from 1.6 to 1.9. The percent of substitution is 27.5 - 31.5 %.

      Helpful?

Reviews

No rating value

Active Filters