Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl cellulose
Synonym(s):
Methyl cellulose
CAS Number:
9004-67-5
Products
Product Category
carbohydrates (10)
polysaccharides (6)
Brand
Sigma-Aldrich (11)
Biological Source
Color
Feature
Physical Form
Agency
Tissue
Quality Segment
Application
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
M0512
Methyl cellulose,
viscosity: 4,000 cP
Expand
View Pricing
M7027
Methyl cellulose,
viscosity: 15 cP, BioReagent, suitable for cell culture
Expand
View Pricing
M0387
Methyl cellulose,
viscosity: 1,500 cP
Expand
View Pricing
M7140
Methyl cellulose,
viscosity: 15 cP
Expand
View Pricing
M6385
Methyl cellulose,
viscosity: 25 cP
Expand
View Pricing
M0262
Methyl cellulose,
27.5-31.5% (Methoxyl content), viscosity: 400 cP
Expand
View Pricing
M0430
Methyl cellulose,
meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Expand
View Pricing
M0555
Methyl cellulose,
26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Expand
View Pricing
94378
Methyl cellulose,
viscosity 3000-5500 mPa.s, 2 % in H
2
O(20 °C)
Expand
View Pricing
64625
Methyl cellulose,
medium viscosity, Methoxyl content 27.5-31.5 %
Expand
View Pricing
64632
Methyl cellulose,
tested according to Ph. Eur.
Expand
View Pricing
64605
Methyl cellulose,
27.5-31.5% methoxyl basis
Expand
View Pricing