Skip to Content
Merck
All Photos(2)

Key Documents

69293

Sigma-Aldrich

Sucralose

≥98.0% (HPLC)

Synonym(s):

1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19Cl3O8
CAS Number:
Molecular Weight:
397.63
Beilstein:
3654410
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D 86.0±2.0°, c = 1 in H2O

sweetness

600 × sucrose

color

white to off-white

useful pH range

6-8 (20 °C, 100 g/L)

mp

114.5  °C ((238.1 °F ) - OECD Test Guideline 102)

solubility

~300 g/L at 25 °C (77 °F)

SMILES string

OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

InChI

1S/C12H19Cl3O8/c13-1-4-7(17)10(20)12(3-14,22-4)23-11-9(19)8(18)6(15)5(2-16)21-11/h4-11,16-20H,1-3H2/t4-,5-,6+,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

BAQAVOSOZGMPRM-QBMZZYIRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.

Biochem/physiol Actions

A synthetic sweet tastant detectable by humans. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells and elicits increased hormonal secretion of glucagon-like peptide-1 and glucose-dependent insulinotrophic peptide.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nimesh Shukla et al.
Archives of biochemistry and biophysics, 639, 38-43 (2017-12-31)
Sucralose is a commonly employed artificial sweetener that appears to destabilize protein native structures. This is in direct contrast to the bio-preservative nature of its natural counterpart, sucrose, which enhances the stability of biomolecules against environmental stress. We have further
Ann-Kristin Eriksson Wiklund et al.
Chemosphere, 86(1), 50-55 (2011-10-01)
The non-calorie sweetener sucralose - sucrose containing three chlorine atoms - is intensively sweet and has become a popular substitute for sugar. Its widespread use, exceptional stability in combination with high water solubility have thus resulted in contamination of recipient
Virender K Sharma et al.
Chemosphere, 87(6), 644-648 (2012-02-22)
The kinetics of the oxidation of sucralose, an emerging contaminant, and related monosaccharides and disaccharides by ferrate(VI) (Fe(VI)) were studied as a function of pH (6.5-10.1) at 25°C. Reducing sugars (glucose, fructose, and maltose) reacted faster with Fe(VI) than did
Christopher B Forsyth et al.
PloS one, 6(12), e28032-e28032 (2011-12-07)
Parkinson's disease (PD) is the second most common neurodegenerative disorder of aging. The pathological hallmark of PD is neuronal inclusions termed Lewy bodies whose main component is alpha-synuclein protein. The finding of these Lewy bodies in the intestinal enteric nerves
Ana I Domingos et al.
Nature neuroscience, 14(12), 1562-1568 (2011-11-15)
We developed an assay for quantifying the reward value of nutrient and used it to analyze the effects of metabolic state and leptin. In this assay, mice chose between two sippers, one of which dispensed water and was coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service