Skip to Content
Merck
All Photos(4)

Documents

HPA038885

Sigma-Aldrich

Anti-YAP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-YAP65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PRKAMLSQMNVTAPTSPPVQQNMMNSASGPLPDGWEQAMTQDGEIYYINHKNKTTSWLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YAP1(10413)

General description

Yes-associated protein 1 (YAP1) is a transcriptional coactivator and has two WW domains (two conserved tryptophans), TID domain (TEA domain family member 1 (TEAD) transcription factor interacting domain), proline rich region, sarcoma homology 3 domain (SH3) binding motif and post synaptic density protein (PSD95), Drosophila disc large tumor suppressor (Dlg1), and zonula occludens-1 protein motif (PDZ). It is expressed in eight isoforms. The gene YAP1 is mapped to human chromosome 11q22.1.

Immunogen

Yes-associated protein 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Yes-associated protein 1 (YAP1) interacts with the src family kinase c-Yes by its SH3 binding motif and with protein 73 using WW motif. YAP promotes cell and tissue growth. YAP1 interacts with Receptor tyrosine-protein kinase (ErbB4) receptor and regulates transcription. YAP is oncogenic and contributes to tumor progression. Phosphorylation of YAP is critical for regulating apoptosis. YAP1 levels are elevated in human cancers. It is highly expressed in hedgehog-associated medulloblastomas. Mutations in YAP1 is also implicated in optic fissure closure defects. Knockdown of YAP1 gene impacts on prostate cancer progression and is considered as potential target for treatment.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST87319

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yap1 phosphorylation by c-Abl is a critical step in selective activation of proapoptotic genes in response to DNA damage
Levy D, et al.
Molecular Cell, 29(3), 350-361 (2008)
JNK phosphorylates Yes-associated protein (YAP) to regulate apoptosis
Tomlinson V, et al.
Cell Death & Disease, 1(2), e29-e29 (2010)
WW domain-containing protein YAP associates with ErbB-4 and acts as a co-transcriptional activator for the carboxy-terminal fragment of ErbB-4 that translocates to the nucleus
Komuro A, et al.
The Journal of Biological Chemistry, 278(35), 33334-33341 (2003)
Inactivation of YAP oncoprotein by the Hippo pathway is involved in cell contact inhibition and tissue growth control
Zhao B, et al.
Genes & Development, 21(21), 2747-2761 (2007)
Identification, basic characterization and evolutionary analysis of differentially spliced mRNA isoforms of human YAP1 gene
Gaffney CJ, et al.
Gene, 509(2), 215-222 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service