Skip to Content
Merck

Y4770

Anti-YAP1 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-YAP, Anti-YAP2, Anti-YAP65, Anti-Yes-associated protein 1, 65kDa

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
10
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using HEK-293T cells co-transfected with human YAP1 and c-Abl

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YAP1(10413)
mouse ... Yap1(22601)
rat ... Yap1(363014)

General description

YAP1 (Yes-associated protein 1, YAP, YAP2, YAP65) is a modular adaptor protein with multiple protein interaction domains, that is based on its interaction with Src-family tyrosine kinase c-Yes. It has a SH3-binding motif, proline-rich amino terminus, a WW domain, a coiled-coil and a PDZ-binding motif at the extreme C-terminus. YAP1 also contains a 14-3-3 interacting motif.

Immunogen

synthetic peptide corresponding to amino acids 442-454 located at the C-terminus of human YAP1. The sequence is identical in mouse, bovine, and dog YAP1.

Application

Anti-YAP1 (C-terminal) antibody is suitable for use in western blot (.5-1 μg/mL) using HEK-293T cells co-transfected with human YAP1 and c-Abl.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Anti-YAP1 (C-terminal) antibody is specific for human YAP1. The product is expected to react with mouse, bovine, and dog YAP1. In immunoblotting, detection of the YAP1 band is specifically inhibited with the immunizing peptide.
YAP1 (Yes-associated protein 1) induces p73-dependent apoptosis through the specific and selective co-activation of p53AIP1, an apoptotic p73 target gene. It stabilizes p73 by preventing Itch-mediated ubiquitination of p73. Upon phosphorylation by Akt at Ser127, 14-3-3 is recruited and stimulates YAP1 localization to the cytoplasm, resulting in loss of co-activator function in the nucleus. Inhibition of Akt potentiates the nuclear re-localization of YAP1 to promote apoptosis by p73.
YAP1 is a WW domain containing protein that functions as a transcriptional co-activator . It may also regulate p73 gene function in response to DNA damage. YAP1 is known to associate with a wide range of transcription factors . Alterations in YAP1 have been associated with tumor development .

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library