Skip to Content
Merck
All Photos(1)

Documents

SAB2501938

Sigma-Aldrich

Anti-YAP1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

YAP, YAP2, YAP65, Yes-associated protein 1, 65 kD, Yes-associated protein 1, 65 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, canine, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... YAP1(10413)

Related Categories

Immunogen

Peptide with sequence C-TYHSRDESTDS, from the internal region of the protein sequence according to NP_006097.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maolan Li et al.
Cancer letters, 355(2), 201-209 (2014-09-15)
The transcriptional coactivator Yes-associated protein 1 (YAP1), a key regulator of cell proliferation and organ size in vertebrates, has been implicated in various malignancies. However, little is known about the expression and biological function of YAP1 in human gallbladder cancer
Masahiro Tsujiura et al.
PloS one, 9(6), e100974-e100974 (2014-06-28)
Yes-associated protein (YAP) is a transcriptional co-activator and regulates cell proliferation and apoptosis. We investigated the clinical and biological significance of YAP in endometrial cancer (EMCA). YAP expression in 150 primary tumor tissues from patients with EMCA was evaluated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service