Skip to Content
Merck
All Photos(3)

Documents

SAB4501060

Sigma-Aldrich

Anti-GAB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GRB2-associated binder 1, GRB2-associated-binding protein 1, growth factor receptor bound protein 2-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 76 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAB1(2549)

Related Categories

General description

Anti-GAB1 Antibody detects endogenous levels of total GAB1 protein.
The GRB2 associated binding protein 1 (GAB1) protein is encoded by the gene mapped to human chromosome 4q31.21. The encoded protein belongs to the Grb2-associated binding protein family. GAB1 protein structure contains pleckstrin homology domain and tyrosine residues. Anti-GAB1 Antibody detects endogenous levels of total GAB1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human GAB1.

Immunogen Range: 644-674

Application

Anti-GAB1 antibody produced in rabbit has been used in proximity ligation assay (1:100) and western blotting.

Biochem/physiol Actions

GRB2 associated binding protein 1 (GAB1) acts as a scaffold protein for protein-protein interaction. It regulates growth factor receptors and enhances signal transduction of multiple intracellular signaling pathways, such as phosphatidylinositol 3-kinase and protein kinase B (PKB/AKT)/ mammalian target of rapamycin (mTOR) signaling pathways. Upregulated expression of GAB1 contributes to the progression of various types of cancer including human oral squamous cell carcinoma (OSCC), breast cancer, colorectal cancer, hepatocellular carcinoma, and ovarian cancer. Therefore, knockdown of the GAB1 gene can be considered as a potential therapeutic strategy for treating breast cancer and OSCC.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethar Al-Husseinawi et al.
Pathology, 51(6), 610-614 (2019-08-25)
The Grb2-associated binding proteins are a family of platform proteins involved in signalling transduction pathways. A major family member, Gab1, modulates the effect of growth factor receptors and intracellular signalling pathways, including the phosphatidylinositol 3-kinase and AKT/mTOR signalling pathways. We
Luyong Xu et al.
Oncology research, 26(4), 617-624 (2017-09-13)
Grb2-associated binder 1 (Gab1) is often aberrant in cancerous cells and tissues, whose alteration is responsible for aggressive phenotypes. In this study, we examined the Gab1 expression in human oral squamous cell carcinoma (OSCC) tissues and investigated the cellular and
Xiao Wang et al.
Journal of experimental & clinical cancer research : CR, 38(1), 27-27 (2019-01-23)
Breast cancer (BCa) remains as the second leading cause of cancer-related death in women worldwide. The majority of the deaths are due to its progression to metastatic BCa. Although Grb2-associated binding protein 1 (Gab1) has been implicated in tumor proliferation
Rui Xu et al.
Genetics and molecular biology, 46(2), e20220277-e20220277 (2023-06-05)
Long non-coding RNA AK001796 was initially identified altered in lung cancer. Recent research showed it could participate in the prognosis of hepatocellular carcinoma (HCC). However, the general biological role of AK001796 and its underlying mechanisms in HCC remain unclear. Here
Microarray analysis of androgenetic and senescent alopecia: comparison of gene expression shows two distinct profiles.
Pratima Karnik et al.
Journal of dermatological science, 72(2), 183-186 (2013-07-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service