Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
FibriCol®, Collagen solution from Bovine hide, 97% Type I with the remainder of Type III collagen, suitable for cell culture
biological source
(Collagen solution from Bovine hide)
Quality Segment
sterility
sterile-filtered
assay
>99% (SDS-PAGE Electrophoresis – Silver Stain, SDS-PAGE)
form
solution
packaging
pkg of 20 mL
technique(s)
cell culture | mammalian: suitable
solubility
water: miscible
shipped in
wet ice
storage temp.
2-8°C
General description
It is ideal for applications where high concentrations of collagen are needed. FibriCol® collagen is provided in a 20 mL volume and is contained in user-friendly packaging for use and storage. FibriCol® is sterile filtered and is supplied as a ready to use solution.
Application
- in the preparation of liposome-loaded co-gels to treat methicillin-resistant Staphylococcus aureus (MRSA) infections in vitro and in vivo[1]
- in the preparation of gelatin-methacrylate (gelMA)-collagen gels to study the effects of stiffness and fiber density on cellular behavior in breast cancer cell lines[2]
- in the preparation of collagen hydrogels to study its effects on macrophage response in partial thickness abdominal wall defect rat models
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| sterility sterile-filtered | sterility sterile-filtered | sterility sterile-filtered | sterility sterile-filtered |
| biological source (Collagen solution from Bovine hide) | biological source bovine skin | biological source rat tail | biological source bovine |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable using and for 3D matrix formation. | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| assay >99% (SDS-PAGE Electrophoresis – Silver Stain, SDS-PAGE) | assay - | assay >95% (SDS-PAGE) | assay 99.9% |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| form solution | form liquid | form liquid | form lyophilized |
Still not finding the right product?
Explore all of our products under FibriCol®
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



