Skip to Content
Merck
All Photos(1)

Key Documents

G9888

Sigma-Aldrich

Glycerol 3-phosphate Oxidase from Aerococcus viridans

lyophilized powder, ≥70 units/mg solid

Synonym(s):

sn-Glycerol 3-phosphate:oxygen 2-oxidoreductase, L-Glycerol 3-phosphate Oxidase, GPO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

specific activity

≥70 units/mg solid

composition

Protein, ≥60% Lowry

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will oxidize 1.0 μmole of L-glycerol 3-phosphate to dihydroxyacetone phosphate with the formation of H2O2 per min at 37°C, at pH 8.1.

Physical form

Lyophilized powder containing sucrose

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Albert Premkumar et al.
Physiologia plantarum, 167(1), 90-110 (2018-11-13)
Hypoxia (oxygen deprivation) causes metabolic disturbances at physiological, biochemical and genetic levels and results in decreased plant growth and development. Phospholipase D (PLD)-mediated signaling was reported for abiotic and biotic stress signaling events in plants. To investigate the participatory role
Tatsuya Kishimoto et al.
Clinica chimica acta; international journal of clinical chemistry, 333(1), 59-67 (2003-06-18)
Several methods for measuring concentrations of lysophosphatidic acid (LPA), a lipid mediator, have been reported to date. However, these methods are not routinely used because most of them require specialized instrument and a complicated protocol. We developed a novel LPA
C S Pundir et al.
Clinical biochemistry, 43(4-5), 467-472 (2009-12-29)
A method is described for construction of an amperometric triglyceride (TG) biosensor using PVA membrane bound enzymes. A mixture of commercial lipase, glycerol kinase, glycerol-3-phosphate oxidase and horseradish peroxidase was co-immobilized onto polyvinyl alcohol (PVA) membrane through glutaraldehyde coupling. The
Shu-Yi Hsu et al.
Sensors (Basel, Switzerland), 10(6), 5758-5773 (2010-01-01)
A single use, disposable iridium-nano particle contained biosensor had been developed for the determination of diglyceride (DG). In this study hydrogen peroxide, formed through the enzymatic breakdown of DG via lipase, glycerol kinase and glycerol 3-phosphate oxidase, was electrochemically oxidized
Shao-Peng Chen et al.
The Analyst, 135(11), 2979-2986 (2010-09-30)
In this paper an enzyme-carrier-based microfluidic chip coupled with a gold nanoband microelectrode as electrochemical detector for Triglyceride (TG) determination was developed by co-immobilized lipase, Glycerokinase (GK) and glycerol-3-phosphate oxidase (GPOx) on chitosan/Fe(3)O(4) composite nanoparticles with a shell-core structure, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service