Skip to Content
Merck
All Photos(1)

Key Documents

G6278

Sigma-Aldrich

Glycerokinase from Escherichia coli

300-600 units/mL, ammonium sulfate suspension

Synonym(s):

ATP:glycerol 3-phosphotransferase, Glycerol Kinase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.77

biological source

Escherichia coli

Quality Level

form

ammonium sulfate suspension

concentration

300-600 units/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Research Area: Cell Signaling
Glycerol kinase (GK) is a member of the FGGY carbohydrate kinase family (ATP: glycerol 3-phosphotransferase), which is responsible for transferring a phosphate group from ATP to glycerol.

Application

Glycerol kinase (glpK) was used to study the effects of pain controlling neuropeptides on human fat cell lipolysis.
Glycerol kinase has been used along with adenosine triphosphate (ATP) for the conversion of glycerol to glycerol-3 phosphate (glycerol-3P) in white adipose tissue (WAT) adipocyte samples by label distribution method. It has also been used along with adenosine triphosphate (ATP) in the derivatization of glycerol to sn-glycerol-3 phosphate (glycerol-3P).

Biochem/physiol Actions

Glycerol kinase (GK), the primary enzyme involved in regulating glycerol uptake and its metabolism, was initially identified in E. coli. This enzyme acts as a catalyst in the Mg2+-ATP-dependent phosphorylation of glycerol to produce G3P, a crucial intermediate in various metabolic pathways, including the synthesis of glycerolipids and triglycerides, as well as glycogenesis, glycolysis, and gluconeogenesis.
GK participates in the glycerol 3-phosphate shuttle and, in conjunction with glycerol 3-phosphate dehydrogenase, converts glycerol into dihydroxyacetone phosphate. It plays a pivotal role in plasma glycerol withdrawal, glycerol utilization by different tissues, and in maintaining carbohydrate homeostasis. GK is positioned at the nexus of lipid and carbohydrate metabolism and may play a potential role in type 2 diabetes mellitus (T2DM).
Glycerol kinase catalyzes tge MgATP-dependent phosphorylation of glycerol to produce sn-glycerol-3-phosphate and is the rate limiting enzyme in the utilization of glycerol. It is also subject to feedback regulation by fructose-1,6-bisphosphate.

Unit Definition

One unit will convert 1.0 μmole of glycerol and ATP to L-α-glycerophosphate and ADP per min at pH 9.8 at 25 °C in a coupled system with PK/LDH.

Physical form

Suspension in 3.1 M (NH4)2SO4 pH 7.3, with 1% BSA and 2% trehalose

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wendy Escobedo-Hinojosa et al.
MethodsX, 8, 101285-101285 (2021-08-27)
Phosphorous-NMR is scarcely employed to evaluate enzyme kinetics of kinase driven monophosphorylations, despite of being a powerful and reliable tool to undoubtedly detect the actual phosphoryl transfer to the targeted substrate. Another advantage is that an external supplementation source of
Ana Cecilia Ho-Palma et al.
Nutrients, 11(3) (2019-03-03)
Under normoxic conditions, adipocytes in primary culture convert huge amounts of glucose to lactate and glycerol. This "wasting" of glucose may help to diminish hyperglycemia. Given the importance of insulin in the metabolism, we have studied how it affects adipocyte
Expression and characterisation of human glycerol kinase: the role of solubilising agents and molecular chaperones
Rani RM, et al.
Bioscience Reports, 43(4), BSR20222258-BSR20222258 (2023)
V van Harmelen et al.
International journal of obesity (2005), 34(8), 1333-1340 (2010-03-17)
Neuropeptides NPFF and NPSF are involved in pain control, acting through the G-protein coupled receptors (GPR)74 (high affinity for NPFF) and GPR147 (equal affinity for NPFF and NPSF). GPR74 also inhibits catecholamine-induced adipocyte lipolysis and regulates fat mass in humans.
Eric Chiles et al.
Analytical biochemistry, 575, 40-43 (2019-04-04)
Glucose and glycerol are important circulating metabolites. Due to poor ionization and/or ion suppression, the liquid chromatography-mass spectrometry (LC-MS) detection of glucose and glycerol presents challenges. Here, we propose an efficient LC-MS method of quantitative glucose and glycerol detection via

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service