Skip to Content
Merck
All Photos(1)

Key Documents

G9145

Sigma-Aldrich

[Leu15]-Gastrin I human

≥95% (HPLC)

Synonym(s):

[Leu15]-HG-17, LHG-17, [15-Leucine]-Gastrin I, human heptadecapeptide, [18-Pyroglutamic acid, 32-leucine]-Big gastrin I-(18-34)-peptide amide, human, [Leu15]-Gastrin I, human, [Leucine15]-Gastrin I heptadecapeptide, [Leucine15]-Little gastrin I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C98H126N20O31
CAS Number:
Molecular Weight:
2080.16
Beilstein:
5227423
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

human

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)C3CCCN3C(=O)CNC(=O)C4CCC(=O)N4)C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(C)C(=O)NC(Cc5ccc(O)cc5)C(=O)NCC(=O)NC(Cc6c[nH]c7ccccc67)C(=O)NC(CC(C)C)C(=O)NC(CC(O)=O)C(=O)NC(Cc8ccccc8)C(N)=O

InChI

1S/C98H126N20O31/c1-49(2)38-68(115-96(147)72(43-55-46-101-60-19-12-10-17-58(55)60)117-98(149)74-20-13-37-118(74)77(122)48-103-86(137)61-25-31-75(120)105-61)93(144)111-66(30-36-82(131)132)92(143)110-65(29-35-81(129)130)91(142)109-64(28-34-80(127)128)90(141)108-63(27-33-79(125)126)89(140)107-62(26-32-78(123)124)88(139)104-51(5)85(136)113-70(41-53-21-23-56(119)24-22-53)87(138)102-47-76(121)106-71(42-54-45-100-59-18-11-9-16-57(54)59)95(146)114-69(39-50(3)4)94(145)116-73(44-83(133)134)97(148)112-67(84(99)135)40-52-14-7-6-8-15-52/h6-12,14-19,21-24,45-46,49-51,61-74,100-101,119H,13,20,25-44,47-48H2,1-5H3,(H2,99,135)(H,102,138)(H,103,137)(H,104,139)(H,105,120)(H,106,121)(H,107,140)(H,108,141)(H,109,142)(H,110,143)(H,111,144)(H,112,148)(H,113,136)(H,114,146)(H,115,147)(H,116,145)(H,117,149)(H,123,124)(H,125,126)(H,127,128)(H,129,130)(H,131,132)(H,133,134)

InChI key

CMVMLPDUAGUTOC-UHFFFAOYSA-N

Gene Information

human ... GAST(2520)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Leu-Asp-Phe-NH2

General description

GAST gene encodes for the protein [Leu15]-Gastrin I in human. In human chromosome, the gene GAST is localized on 17q21.2. G cells in the stomach antrum produces the precursor of [Leu15]-Gastrin I, progastrin. Progastrin undergoes cleavage and processing to yield gastrin, which is trophic for the entire gastrointestinal epithelium. Gastrin is essential for the growth of the digestive system and stimulates the production of gastric acid by parietal cells. Gastrin exerts its function through G-protein-coupled receptor called the cholecystokinin (CCK) or CCK-B receptor (CCK-BR). Gastrin release is stimulated by food, especially protein diet and is inhibited by very low pH. Atrophic gastritis, a Helicobacter pylori infection and long-term administration of proton pump inhibitors infection may cause overexpression of gastrin. Gastric adenocarcinoma show high levels of gastrin.

Application

Gastrin has been used:
  • as a medium supplement establish enteroid cultures
  • as a supplement to standard Dulbecco′s Modified Eagle Medium (DMEM) culture medium, human minigut medium to establish the growth of crypts.

Biochem/physiol Actions

Gastrin I analog with full biological activity and improved stability in aqueous solutions. Leu15 replaces Met15, that is readily oxidized resulting in loss of biological activity.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Establishment of human epithelial enteroids and colonoids from whole tissue and biopsy
Mahe MM, et al.
Journal of Visualized Experiments, (97), e52483-e52483 (2015)
D Chen et al.
Scandinavian journal of gastroenterology, 31(4), 404-410 (1996-04-01)
Gastrin is thought to stimulate growth of the pancreas via gastrin/cholecystokinin (CCK)-B-type receptors. The aim of the present study was to examine the trophic response of the pancreas to exogenous gastrin or to hypergastrinemia of endogenous origin and to hypogastrinemia
C M Zhao et al.
Cell and tissue research, 291(2), 315-323 (1998-03-28)
Ageing cells, especially post-mitotic cells, are known to accumulate pigments, i.e. highly electron-dense material, referred to as ceroid or lipofuscin. This material is formed as a consequence of autophagocytosis and peroxidation of the products undergoing degradation. The present study describes
Gastrin and gastric cancer
Smith JP, et al.
Cellular and molecular gastroenterology and hepatology, 4(1), 75-83 (2017)
Thrombin modifies growth, proliferation and apoptosis of human colon organoids: a protease-activated receptor 1-and protease-activated receptor 4-dependent mechanism
Sebert M, et al.
British Journal of Pharmacology, 175(18), 3656-3668 (2018)

Articles

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service