Skip to Content
Merck

SML0788

A 83-01

≥98% (HPLC), powder, TGF-β RI kinase inhibitor

Synonym(s):

3-(6-Methyl-2-pyridinyl)-N-phenyl-4-(4-quinolinyl)-1H-pyrazole-1-carbothioamide, A-83-01, A83-01

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H19N5S
CAS Number:
Molecular Weight:
421.52
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

A 83-01, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

S=C([n]2nc(c(c2)c4c5c(ncc4)cccc5)c3nc(ccc3)C)Nc1ccccc1

InChI

1S/C25H19N5S/c1-17-8-7-13-23(27-17)24-21(19-14-15-26-22-12-6-5-11-20(19)22)16-30(29-24)25(31)28-18-9-3-2-4-10-18/h2-16H,1H3,(H,28,31)

InChI key

HIJMSZGHKQPPJS-UHFFFAOYSA-N

General description

A-83-01 may effectively prevent burn wound contraction without impeding wound closure, due to its ability to inhibit the transforming growth factor-β (TGF-β)-induced rise in myofibroblast population. Additionally, it can hinder TGF-β1-dependent cancer metastasis by suppressing epithelial-mesenchymal transition (EMT) in animals.

Application

A 83-01 has been used:
  • in the culture medium for organoid formation from dissociated tumor cells,
  • as a component in Dulbecco′s modified Eagle medium/nutrient mixture F-12 (DMEM/F12) for culturing human epidermal stem cells (EpSCs)

A 83-01 has been used as an inhibitor of transforming growth factor β kinase type 1 receptor.

Biochem/physiol Actions

A 83-01 is a TGFβ kinase/activin receptor-like kinase (ALK 5) inhibitor (IC50=12 nM) that prevents phosphorylation of Smad2/3 and inhibits growth induced by TGFβ. A 83-01 blocks phosphorylation of Smad2 and inhibits TGF-β-induced epithelial-to-mesenchymal transition. Also, A 83-01 inhibits the transcriptional activity induced by TGFβ type I receptor ALK-5, activin type IB receptor ALK-4 and nodal type I receptor ALK-7. A-83-01 induces an expansion of neonatal Nkx2.5-eGFP (+) cells.
A-83-01 is a TGFβ kinase/activin receptor-like kinase (ALK 5) inhibitor.


Still not finding the right product?

Explore all of our products under A 83-01


Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library