Skip to Content
Merck

A0737

N-acetylcysteine amide

≥98% (HPLC), lyophilized powder, antioxidant

Synonym(s):

(R)- 2-(Acetylamino)-3-mercapto-Propanamide, AD4, N-Acetyl-L-cysteinamide, NACA, acetylcysteinamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H10N2O2S
CAS Number:
Molecular Weight:
162.21
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Assay:
≥98% (HPLC)
Form:
lyophilized powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-acetylcysteine amide, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

lyophilized powder

storage condition

desiccated

color

white to off-white

solubility

H2O: ≥20 mg/mL, DMSO: >40 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](CS)C(N)=O

InChI

1S/C5H10N2O2S/c1-3(8)7-4(2-10)5(6)9/h4,10H,2H2,1H3,(H2,6,9)(H,7,8)/t4-/m0/s1

InChI key

UJCHIZDEQZMODR-BYPYZUCNSA-N

Application

N-acetylcysteine amide has been used:
  • to protect retinal pigment epithelium (RPE) from oxidative stress
  • to show that increased expression of cell adhesion molecule 4 (CADM4) in oligodendrocytes inhibits myelination
  • to protect cells from apoptosis induced by shikonin plus erlotinib/gefitinib
  • as a component in various culture media

as an antioxidant to reduce oxidative stress in brains of homozygous Drosophila spermine synthase (dSms e/e) mutant flies and melanoma cells.

Biochem/physiol Actions

N-acetylcysteine amide is a membrane penetrating antioxidant with antiinflamatory activity through regulation of activation of NF-κB and HIF-1α as well as modulation of ROS.
N-acetylcysteine amide is a membrane penetrating antioxidant with antiinflamatory activity through regulation of activation of NF-κB and HIF-1α as well as modulation of ROS. The compound readily crosses cell membranes, replenishes intracellular GSH, and defends the cell from oxidative stress. In contrast to DTT, AD4 is able to directly reduce intracellularl GSSG to GSH without the involvement of glutathione peroxidase. Such direct thiol exchange might have a protective effect. This compound has a potential in research and exploration for treatment of neurodegeneration, radiation exposure, and other oxidation-mediated disorders.

Other Notes

This product is hydroscopic and air sensitive.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library