Skip to Content
Merck
All Photos(1)

Key Documents

G8507

Sigma-Aldrich

α-Galactosidase from green coffee beans

ammonium sulfate suspension, ≥9 units/mg protein

Synonym(s):

alpha-Galactosidase, α-D-Galactoside galactohydrolase, Melibiase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

ammonium sulfate suspension

Quality Level

specific activity

≥9 units/mg protein

storage temp.

2-8°C

General description

α-Galactosidase from green coffee beans is encoded by the GLA gene.

Application

α-Galactosidase from green coffee beans has been used to assess the specificity of anti- α-Galactosidase antibodies. It has also been used in inhibition assay to assess N-guanidino derivatives of 1,5-dideoxy-1,5-imino-d-xylitol activity.

Biochem/physiol Actions

α-Galactosidase from green coffee beans cleaves the terminal α-galactose from the oligosaccharides. α-Galactosidase deficiency is associated with Fabry disease.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl α-D-galactoside to p-nitrophenol and D-galactose per min at pH 6.5 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 solution, pH 6.0, containing BSA

inhibitor

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pig xenogeneic antigen modification with green coffee bean alpha-galactosidase
Luo Y, et al.
Xenotransplantation, 6(4), 238-248 (1999)
A prophylactic alpha-Gal-based glycovaccine effectively protects against murine acute Chagas disease
Portillo S, etal.
NPJ vaccines, 4(1), 13-13 (2019)
Amblyomma sculptum tick saliva: alpha-Gal identification, antibody response and possible association with red meat allergy in Brazil
Araujo RN, et al.
International Journal For Parasitology, 46(3), 213-220 (2016)
Structure-function relationships in alpha-galactosidase A
Garman SC
Acta Paediatrica (Oslo, Norway : 1992), 96, 6-16 (2007)
Orthoester functionalized N-guanidino derivatives of 1, 5-dideoxy-1, 5-imino-d-xylitol as pH-responsive inhibitors of beta-glucocerebrosidase
Sevvsek A, et al.
MedChemComm, 8(11), 2050-2054 (2017)

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service