Skip to Content

N0877

4-Nitrophenyl α-D-galactopyranoside

α-glucosidase substrate, chromogenic, ≥99% (TLC), powder

Synonym(s):

p-Nitrophenyl α-D-galactopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
231-343-6
MDL number:
Beilstein/REAXYS Number:
92214
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

4-Nitrophenyl α-D-galactopyranoside, α-galactosidase substrate

Quality Segment

assay

≥99% (TLC)

form

powder

solubility

methanol: 100 mg/mL, clear to very slightly hazy

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9+,10+,11-,12+/m1/s1

InChI key

IFBHRQDFSNCLOZ-IIRVCBMXSA-N

Application

4-Nitrophenyl α-D-galactopyranoside has been used as a substrate to determine α-galactosidase activity.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library