Skip to Content
Merck

Skip To

P6635

Phosphorylase b from rabbit muscle

lyophilized powder, ≥20 units/mg protein, 2× crystallization

Phosphorylase b from rabbit muscle

Synonym(s):

α-Glucan Phosphorylase, 1,4-α-D-Glucan:orthophosphate α-D-glucosyltransferase, Glycogen Phosphorylase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.77
EC Number:
232-737-0
MDL number:
EC Number:
Specific activity:
≥20 units/mg protein
Biological source:
rabbit muscle
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit muscle

Quality Segment

form

lyophilized powder

specific activity

≥20 units/mg protein

mol wt

97,200 Da by calculation

purified by

2× crystallization

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

technique(s)

mass spectrometry (MS): suitable

impurities

~0.01 μmol/mg protein 5′-AMP (This low level will not interfere with phosphorylase and phosphorylase kinase assays.)

UniProt accession no.

foreign activity

phosphoglucomutase ≤1.0%, phosphorylase a ≤10%, phosphorylase kinase ≤0.5%, phosphorylase phosphatase, debrancher enzyme, AMPase and ATPase ≤0.1%

storage temp.

−20°C

General description

Research area: Cell Signaling

Glycogen phosphorylase (PG), a specialized complex allosteric enzyme[1] has an evolutionarily conserved gene sequence.[2] GP contains a family of three isozymes such as muscle GP (mGP), liver GP (lGP), and brain GP (bGP) in humans.[1]

Application

Phosphorylase b from rabbit muscle has been used:
  • in the calibration of Sepharose C1-6B columns while studying the molecular weight of methylamine dehydrogenase subunits[3]
  • in ion mobility-mass spectrometry studies of phosphorylase B ions that have been generated with supercharging reagents, in the charge-reducing buffer[4]
  • for the preparation of p32 labeled phosphorylase A using phosphorylase kinase and [32P]ATP[5]
  • in phosphorylase phosphatase assay[6]
  • in enzyme assay as a positive control to ensure the reaction system for the activity determination was adopted[7]

Biochem/physiol Actions

Phosphorylase b is a non-active form and is present in resting muscles. Phosphorylase b kinase activity increases significantly when the Mg2+:ATP ratio exceeds 1. The breakdown of ATP during muscle contraction is thought to trigger in vivo conversion of phosphorylase b into a[8]. Phosphorylase b is activated by inosine monophosphate[9]. Glycogen phosphorylase (PG) enzyme plays a vital role in the first step of glycogenolysis. In the initial stage of glycogenolysis, glycogen phosphorylase(GP) breaks α-1,4- -glycosidic bonds, releasing the glucose-1-phosphate (G1P)molecule.[2] When incubated with the proper concentrations of glucose-1-phosphate without the addition of primer, rabbit muscle phosphorylase B was found to be capable of forming protein-bound alpha-1,4 glucosyl chains.[10]

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing lactose, 5′-AMP, and Mg(OAc)2 (10 μmole per 100 mg protein)

Other Notes

One unit will form 1.0 μmole of α-D-glucose 1-phosphate from glycogen and orthophosphate in the presence of 5′-AMP, per min at pH 6.8 at 30 °C measured in a system containing phosphoglucomutase, NADP, and glucose 6-phosphate dehydrogenase. (One μmolar unit is equivalent to approx. 45 Cori units.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4649P1261P3397
specific activity

≥20 units/mg protein

specific activity

-

specific activity

20-30 units/mg protein

specific activity

≥100 units/mg protein

biological source

rabbit muscle

biological source

-

biological source

rabbit muscle

biological source

rabbit muscle

technique(s)

mass spectrometry (MS): suitable

technique(s)

-

technique(s)

-

technique(s)

activity assay: suitable

form

lyophilized powder

form

powder

form

lyophilized powder

form

ammonium sulfate suspension

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

storage condition

-

storage condition

-

storage condition

(Tightly closed)

mol wt

97,200 Da by calculation

mol wt

observed mol wt ~97 kDa

mol wt

-

mol wt

-


Still not finding the right product?

Explore all of our products under Phosphorylase b from rabbit muscle


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Has the amino acid sequence of phosphorylase from rabbit muscle been published?

    1 answer
    1. The sequence has been published. The amino acid composition is:Asp5l, Asn45, Thr35, Ser29, Glu64, Gln31, Pro36, Gly48, Ala63, Cys9, Val62, Met2l, Ile49, Leu79, Tyr36, Phe38, Trp12, Lys48, HiS22, and Arg63. Reference for the sequence and amino acid composition: K. Titani et al., Proc. Nat. Acad. Sci., USA, 74, 4762-4766 (1977) (see attached pdf).Product P1261 is phosphorylase a, which is the tetrameric form. Products P4649 and P6635 are phosphorylase b, the dimeric  form. The sequence information of the subunit would be the same for both forms.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

Active Filters

  1. Pittsburgh, PA
    • Review 1
    • Votes 2
    2 out of 5 stars.

    Wrong protein

    Hi, we ran Mass Spec analysis of the protein band cut from SDS-PAGE running product sold under Cat#P6635, there is no match with any of the UniProt accession numbers listed for this product. The protein band also runs well above 100 kDa marker which is more than expected MW.

    Helpful?

    1. Response from Merck KGaA:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to contact us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.