Skip to Content
Merck
All Photos(4)

Key Documents

D7128

Sigma-Aldrich

L-Dihydroorotic acid

≥99%

Synonym(s):

2,6-Dioxohexahydro-4-pyrimidinecarboxylic acid, L-Hydroorotic acid, Dihydro-L-orotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Quality Level

Assay

≥99%

form

powder

mp

254-255 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1CC(=O)NC(=O)N1

InChI

1S/C5H6N2O4/c8-3-1-2(4(9)10)6-5(11)7-3/h2H,1H2,(H,9,10)(H2,6,7,8,11)/t2-/m0/s1

InChI key

UFIVEPVSAGBUSI-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Dihydroorotic acid has been used as a substrate in dihydroorotate dehydrogenase (DHODH) assay.

Biochem/physiol Actions

L-Dihydroorotic acid (DHO) serves as a substrate for dihydroorotate dehydrogenase (DHODH), an enzyme in the de novo synthesis of pyrimidine. Inhibition of DHOH by its inhibitors causes a large accumulation of upstream metabolite DHO and a reduction in the uridine levels. Therefore, DHO and uridine can be used as biomarkers for pyrimidine synthesis for the clinical development of DHOH inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Yin et al.
Journal of pharmaceutical and biomedical analysis, 192, 113669-113669 (2020-10-30)
Uridine and L-dihydroorotate (DHO) are important intermediates of de novo as well as salvage pathways for the biosynthesis of pyrimidines, which are the building blocks of nucleic acids - DNA and RNA. These metabolites are known to be significant biomarkers
E Takashima et al.
Parasitology international, 50(4), 273-278 (2001-11-24)
Using N2 cavitation, we established a protocol to prepare the active mitochondria from Plasmodium falciparum showing a higher succinate dehydrogenase activity than previously reported and a dihydroorotate-dependent respiration. The fact that fumarate partially inhibited the dihydroorotate dependent respiration suggests that
Mark A Anderson et al.
Biochemistry, 45(23), 7132-7139 (2006-06-07)
In the pyrimidine biosynthetic pathway, N-carbamyl-L-aspartate (CA-asp) is converted to L-dihydroorotate (DHO) by dihydroorotase (DHOase). The mechanism of this important reaction was probed using primary and secondary 15N and 13C isotope effects on the ring opening of DHO using isotope
Juliana Cheleski et al.
Analytical biochemistry, 399(1), 13-22 (2009-11-26)
Trypanosoma cruzi dihydroorotate dehydrogenase (TcDHODH) catalyzes the oxidation of l-dihydroorotate to orotate with concomitant reduction of fumarate to succinate in the de novo pyrimidine biosynthetic pathway. Based on the important need to characterize catalytic mechanism of TcDHODH, we have tailored
Yihan Wu et al.
Cell chemical biology, 24(12), 1437-1444 (2017-10-17)
Microbial natural products are genetically encoded by dedicated biosynthetic gene clusters (BGCs). A given BGC usually produces a family of related compounds that share a core but contain variable substituents. Though common, the reasons underlying this divergent biosynthesis are in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service