Skip to Content
Merck

Skip To

C8081

Coenzyme Q2

≥90% (HPLC), liquid, electron carrier

Synonym(s):

2,3-Dimethoxy-5-methyl-6-geranyl-1,4-benzoquinone, Ubiquinone-10

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H26O4
CAS Number:
Molecular Weight:
318.41
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥90%
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Coenzyme Q2, ≥90%

assay

≥90%

form

liquid

storage temp.

−20°C

SMILES string

CC(C(C(OC)=C1OC)=O)=C(C/C=C(C)/CCC=C(C)C)C1=O

InChI

1S/C19H26O4/c1-12(2)8-7-9-13(3)10-11-15-14(4)16(20)18(22-5)19(23-6)17(15)21/h8,10H,7,9,11H2,1-6H3/b13-10+

InChI key

SQQWBSBBCSFQGC-JLHYYAGUSA-N

Biochem/physiol Actions

Acts as an electron carrier and antioxidant in humans

Other Notes

Non-naturally occurring analog of coenzyme Q10.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C9538C7956D7911
form

liquid

form

powder

form

liquid

form

liquid

assay

≥90%

assay

≥98% (HPLC)

assay

≥95%

assay

≥97% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters