Skip to Content
Merck

D7628

Dihydrouracil

powder

Synonym(s):

5,6-Dihydro-2,4-dihydroxypyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352005
EC Number:
207-982-1
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

mp

279-281 °C (lit.)

SMILES string

O=C1CCNC(=O)N1

InChI

1S/C4H6N2O2/c7-3-1-2-5-4(8)6-3/h1-2H2,(H2,5,6,7,8)

InChI key

OIVLITBTBDPEFK-UHFFFAOYSA-N

General description

Dihydrouracil (DiHU) is a minor base found in transfer ribonucleic acid (tRNA). It is similar to uracil with the only exception that the C5-C6 bond is saturated. It crystallized in the monoclinic system with space group P21/C. Its crystalline structure has been analyzed. Its generation from L-cysteine and uracil via photochemical addition has been described.

Application

Dihydrouracil has been used as a standard for ureido group in the colorimentric assay of transfer ribonucleic acid (tRNA).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ryota Hidese et al.
Journal of bacteriology, 193(4), 989-993 (2010-12-21)
The reductive pyrimidine catabolic pathway is absent in Escherichia coli. However, the bacterium contains an enzyme homologous to mammalian dihydropyrimidine dehydrogenase. Here, we show that E. coli dihydropyrimidine dehydrogenase is the first member of a novel NADH-dependent subclass of iron-sulfur
Stereochemistry of nucleic acids and their constituents. VI. The crystal structure and conformation of dihydrouracil: a minor base of transfer-ribonucleic acid.
Rohrer DC and Sundaralingam M.
Acta Crystallographica Section B, Structural Science, 26(5), 546-553 (1970)
Teresa Di Desidero et al.
Investigational new drugs, 36(4), 709-714 (2018-03-01)
The aim of the present study was to assess the pharmacokinetics (PK) of metronomic capecitabine and its metabolites in a population of refractory metastatic colorectal cancer (mCRC) patients. Thirty-four patients (M/F, 22/12) with a diagnosis of mCRC received capecitabine 800 mg



Global Trade Item Number

SKUGTIN
D7628-5G04061833588611