Skip to Content
Merck

Skip To

M1882

Myoglobin from equine heart

≥90% (SDS-PAGE), essentially salt-free, lyophilized powder

1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

Myoglobin from horse heart

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
309-705-0
NACRES:
NA.61
MDL number:
Form:
essentially salt-free, lyophilized powder
Assay:
≥90% (SDS-PAGE)
Biological source:
equine heart
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

equine heart

Quality Segment

assay

≥90% (SDS-PAGE)

form

essentially salt-free, lyophilized powder

Iron content

≥0.20%

technique(s)

MALDI-MS: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

horse ... MB(100054434)

Application

Myoglobin from equine heart is suitable for use in:
  • spectral measurements in Beckman DU-50 or Gilford 2400 spectrophotometer
  • the secondary structure analysis of proteins in H2O solution using single-pass attenuated total reflection Fourier transform infrared (ATR-FT-IR) microscopy
  • the calibration of the mass scale at a concentration of 2 pmol/μL in Electrospray mass spectrometry
  • a study to investigate on-line single droplet deposition for MALDI mass spectrometry
  • a study to examine protein adsorption in fused-silica and polyacrylamide-coated capillaries

Biochem/physiol Actions

Myoglobin is a mobile carrier of oxygen that is developed in red muscle and heart cells. This happens as a response to elevated demand for oxygen during exercise, and transports oxygen from the sarcolemma to the mitochondria of vertebrate heart and red muscle cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M063092498C2867
description

≥90% (SDS-PAGE), essentially salt-free, lyophilized powder

description

95-100%, essentially salt-free, lyophilized powder

description

from bovine heart, pkg of 500 g, suitable for microbiology

description

BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture

assay

≥90% (SDS-PAGE)

assay

95-100%

assay

-

assay

≥99% (SDS-PAGE)

biological source

equine heart

biological source

equine skeletal muscle

biological source

bovine heart

biological source

horse heart

technique(s)

MALDI-MS: suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

form

essentially salt-free, lyophilized powder

form

essentially salt-free, lyophilized powder

form

powder

form

powder

UniProt accession no.

P68082

UniProt accession no.

P68082

UniProt accession no.

-

UniProt accession no.

P00004

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Myoglobin from equine heart


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–9 of 9 Questions  
  1. Do you have the sequence for Product M1882, Myoglobin from equine heart?

    1 answer
    1. This product has not been specifically sequenced. However, according to the Uniprot accession number P68082, equine myoglobin has the following sequence:

      MGLSDGEWQQVLNVWGKVEADIAGHGQEVLIRLFTGHPETLEKFDKFKHLKTEAEMKASE DLKKHGTVVLTALGGILKKKGHHEAELKPLAQSHATKHKIPIKYLEFISDAIIHVLHSKHPGDFGADAQGAMTKALELFRNDIAAKYKELGFQG

      Please see the link below to review additional information available at UniProt:

      https://www.uniprot.org/uniprotkb/P68082/entry

      Helpful?

  2. I would like to consult you about the valence state of the ion in MB. Is it 2+ or 3+?

    1 answer
    1. This product is native myoglobin isolated from equine heart and is probably a mixture of oxidized myoglobin (metmyoglobin, Fe3+) and reduced myoglobin (Fe2+). The percentages of oxidized or reduced myoglobin in the product are not determined.

      Helpful?

  3. Was this product recombinantly expressed or was it isolated directly from the heart of the horse ?

    1 answer
    1. This product is native myoglobin isolated from equine heart.

      Helpful?

  4. Is equine myoglobin (M1882) detectable and quantifiable by common laboratory tests (ECL) for the detection of human myoglobin?

    1 answer
    1. This item has not been tested in-house for detectability and quantifiability by common laboratory tests. The specifications for this item do not include testing by these methods.

      Helpful?

  5. Is Product M1882, Myoglobin from equine heart, metmyoglobin?

    1 answer
    1. The M1882 is a mixture, but we have not analyzed the percentages of the oxidized myoglobin or the reduced myoglobin in the product.

      Helpful?

  6. Is Product M1882, Myoglobin from equine heart, oxidized?

    1 answer
    1. Yes, it is oxidized.

      Helpful?

  7. Which has more affinity for oxygen, hemoglobin or myoglobin?

    1 answer
    1. The affinity of myoglobin for oxygen is higher than that of hemoglobin.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  9. What is the molecular weight of Product M1882, Myoglobin from equine heart?

    1 answer
    1. Based on the amino acid sequence (16,950) plus the heme group (616), the molecular weight is approximately 17,600 g/mol.

      Helpful?

Reviews

Active Filters

  1. New York
    • Review 1
    • Votes 0
    4 out of 5 stars.

    Is the myoglobin from equine heart dispersive in water?
    Or what are the solvent where it is dispersive.

    Helpful?

    1. Response from Merck KGaA:

      Thank you for your review. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.