Skip to Content
Merck
All Photos(2)

Key Documents

D4913

Sigma-Aldrich

2′-Deoxycytidine 5′-triphosphate disodium salt

100 mM (pH 7)

Synonym(s):

2′-Deoxycytidine-5′-triphosphoric acid disodium salt, dCTP-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3Na2O13P3
CAS Number:
Molecular Weight:
511.12
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.52
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

liquid

concentration

100 mM (pH 7)

color

colorless

foreign activity

DNase, RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O2

InChI

1S/C9H16N3O13P3.2Na/c10-7-1-2-12(9(14)11-7)8-3-5(13)6(23-8)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;/h1-2,5-6,8,13H,3-4H2,(H,18,19)(H,20,21)(H2,10,11,14)(H2,15,16,17);;/q;2*+1/p-2/t5-,6+,8+;;/m0../s1

InChI key

ABWVCNMFYVEBIB-CDNBRZBRSA-L

Looking for similar products? Visit Product Comparison Guide

General description

2′-Deoxycytidine 5′-triphosphate (dCTP) is a cytidine analog.[1] It is composed of a nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar bound to a chain of three phosphate residues. dCTP is utilized by cells for DNA polymerase catalyzed synthesis of DNA.[2]

Application

2′-Deoxycytidine 5′-triphosphate disodium salt has been used:
  • in reverse transcription polymerase chain reaction (RT -PCR).[3]
  • to study the mutations in HBB gene via amplification refractory mutation system (ARMS)-PCR[4]
  • to C-tail the end-repaired DNA[5]
2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingyue He et al.
Methods in molecular biology (Clifton, N.J.), 484, 193-205 (2008-07-02)
Ribosome display is a powerful method for selection of single-chain antibodies in vitro. It operates through the formation of libraries of antibody-ribosome-mRNA complexes that are selected on immobilized antigen, followed by recovery of the genetic information from the mRNA by
Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, C-C (1989)
Nicole Bertoletti et al.
Protein science : a publication of the Protein Society, 28(9), 1664-1675 (2019-07-14)
The retrovirus HIV-1 has been a major health issue since its discovery in the early 80s. In 2017, over 37 million people were infected with HIV-1, of which 1.8 million were new infections that year. Currently, the most successful treatment
Analysis of beta globin gene mutations in Diyarbakir
Tekecs S, et al.
Turkish Journal of Biology (2021)
Joyce E Karlinsey et al.
Cell host & microbe, 26(3), 426-434 (2019-08-27)
Salmonella enterica serovar Typhi causes typhoid fever only in humans. Murine infection with S. Typhimurium is used as a typhoid model, but its relevance to human typhoid is limited. Non-obese diabetic-scid IL2rγnull mice engrafted with human hematopoietic stem cells (hu-SRC-SCID)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service