Skip to Content
Merck

DNTP100A

Deoxynucleotide Set, 100 mM

Individual dNTPs for routine PCR; 1 mL each

Synonym(s):

PCR dNTPs, dNTP set, dNTPs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.52
UNSPSC Code:
41106305
Form:
liquid
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

concentration

100 mM

technique(s)

PCR: suitable

color

colorless

shipped in

dry ice

storage temp.

−20°C

General description

Deoxynucleotide triphosphates (dNTPs) are negatively charged organic molecules, which contains nucleobase bound to deoxyribose. This dNTPs set includes 1.0 mL each of 100mM aqueous solutions of four deoxyribonucleotides namely deoxyribose adenosine triphosphate (dATP), deoxyribose cytidine triphosphate (dCTP), deoxyribose guanosine triphosphate (dGTP), and deoxyribose thymidine triphosphate (dTTP) in separate tubes for convenience and easy PCR reaction setup.

Application

Deoxynucleotide set, 100 mM has been used in:
  • the amplification of DNA in polymerase chain reaction (PCR)
  • quantitative PCR (qPCR)
  • multiplex PCR

Biochem/physiol Actions

Deoxynucleotide triphosphates (dNTPs) are building blocks of nucleic acid molecules and are acidic in nature. They act as substrates for DNA polymerizing enzymes. dNTPs are essential components of PCR master mixes as it is necessary for DNA amplification reaction. Optimal levels of dNTPs ensure proper chromosomal replication and in turn maintain genome stability.

Features and Benefits

  • Provided as 4 separate tubes of dNTPs
  • Allows optimization of individual dNTP concentrations

Packaging

1.0 mL each of 100 mM dATP, dCTP, dGTP and dTTP


Still not finding the right product?

Explore all of our products under Deoxynucleotide Set, 100 mM


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library