Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 100 mM, pH 7
assay
≥99%
Quality Segment
form
liquid
concentration
100 mM
technique(s)
whole genome amplification: suitable
color
colorless
pH
7
foreign activity
DNase, RNase, none detected
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Na+].Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP(O)(=O)OP(O)([O-])=O)O3
InChI
1S/C10H16N5O12P3.Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(25-7)2-24-29(20,21)27-30(22,23)26-28(17,18)19;/h3-7,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);/q;+1/p-1/t5-,6+,7+;/m0./s1
InChI key
YJWCICGGRLOGEH-VWZUFWLJSA-M
General description
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description 100 mM, pH 7 | description =98%, solution, 100 mM, suitable for DNA sequencing, suitable for DNA amplification | description 10 mM | description 100 mM (pH 7) |
| Quality Level 300 | Quality Level - | Quality Level 200 | Quality Level 200 |
| form liquid | form solution | form liquid | form liquid |
| assay ≥99% | assay 98% | assay ≥99% | assay ≥99% |
| color colorless | color - | color colorless | color colorless |
| shipped in dry ice | shipped in - | shipped in dry ice | shipped in dry ice |
| concentration 100 mM | concentration 100 mM | concentration 10 mM | concentration 100 mM (pH 7) |
Still not finding the right product?
Explore all of our products under 2′-Deoxyadenosine 5′-triphosphate sodium salt solution
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

