Skip to Content

DNTP100

Deoxynucleotide Set, 100 mM

Individual dNTPs for routine PCR; 0.25 mL each

Synonym(s):

DNTP Set for PCR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.52
UNSPSC Code:
41106305
Form:
liquid
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

feature

PCR Additives

concentration

100 mM

technique(s)

PCR: suitable, whole genome amplification: suitable

color

colorless

shipped in

dry ice

storage temp.

−20°C

General description

Deoxynucleotide triphosphates (dNTPs) are organic molecules, which contains nucleobase bound to deoxyribose. This dNTPs set includes 0.25 mL each of 100mM aqueous solutions of four deoxyribonucleotides, deoxyribose adenosine triphosphate (dATP), deoxyribose cytidine triphosphate (dCTP), deoxyribose guanosine triphosphate (dGTP), and deoxyribose thymidine triphosphate (dTTP) in separate tubes for convenience and easy PCR reaction setup.

Application

Deoxynucleotide Set, 100 mM has been used as a component of the reaction mixture for polymerase chain reaction (PCR).It has also been used as a component of the reaction mixture for amplification refractory mutation system polymerase chain reaction (ARMS-PCR) to genotype single-nucleotide polymorphisms (SNPs).
Deoxynucleotide Set, 100 mM has been used:
  • to biotinylate λ-DNA for magnetic tweezers measurements
  • in RAPD PCR
  • in FISH probe generation and labeling
  • in PCR amplification of genomic DNA
  • in multiplex PCR
  • in first-strand cDNA synthesis
  • in TUNEL labeling
  • in RT-PCR
  • as a component of the reaction mixture for polymerase chain reaction (PCR)

Biochem/physiol Actions

Deoxynucleotide triphosphates (dNTPs) are substrates for DNA polymerizing enzymes. They act as building blocks of nucleic acid molecules and are acidic in nature. dNTPs are key components of PCR master mixes and are highly essential to amplify DNA. Optimal levels of dNTPs ensure proper chromosomal replication and in turn maintain genome stability.

Features and Benefits

  • Provided as 4 separate tubes of dNTPs.
  • Allows optimization of individual dNTP concentrations.

Packaging

0.25 mL each of 100 mM dATP, dCTP, dGTP and dTTP


Still not finding the right product?

Explore all of our products under Deoxynucleotide Set, 100 mM


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library