Skip to Content
Merck
All Photos(1)

Key Documents

D2906

Sigma-Aldrich

DMEM/F-12

With ʟ-glutamine, sodium pyruvate and HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture

Synonym(s):

DME/Nutrient Mixture F-12 Ham, DMEM Hams F12, DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: no
L-glutamine: yes
phenol red: no
sodium pyruvate: 0.055 g/L
glucose: 3.15 g/L
HEPES: 15 mM

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham is suitable for the growth of a wide range of cell types in low serum conditions. The removal of phenol red is often preferred by investigators working with clonal and stem cell cultures.

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham has been used to culture:

  • Hep-2 cells
  • mouse fibroblast L‐cells
  • L‐ Wnt family member 3A (Wnt3a) cells
  • primary buffalo embryonic fibroblast (BEF)

Quantity

Formulated to contain 15.6 grams of powder per liter of medium.

Reconstitution

Supplement with 1.2 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Zhang et al.
Osteoarthritis and cartilage, 26(6), 824-833 (2018-03-20)
Adipokines play roles in the pathogenesis of osteoarthritis (OA). Fatty acid binding protein 4 (FABP4) is a novel adipokine that is closely associated with obesity and metabolic diseases. The aim of this study was to discover the potential role of
Nereo Kalebic et al.
EMBO reports, 17(3), 338-348 (2016-01-14)
We have applied the CRISPR/Cas9 system in vivo to disrupt gene expression in neural stem cells in the developing mammalian brain. Two days after in utero electroporation of a single plasmid encoding Cas9 and an appropriate guide RNA (gRNA) into
Eric M Erkenbrack et al.
PLoS biology, 16(8), e2005594-e2005594 (2018-08-25)
Among animal species, cell types vary greatly in terms of number and kind. The number of cell types found within an organism differs considerably between species, and cell type diversity is a significant contributor to differences in organismal structure and
Tao Fu et al.
FEBS letters, 593(15), 2040-2050 (2019-06-04)
Oncostatin M (OSM) is a member of the interleukin-6 (IL-6) family, which functions in embryo implantation and decidualization. The expression, function and regulation of Osm in mouse uteri during early pregnancy remain unknown. We show that Osm is mainly expressed
Peter S B Finnie et al.
Current biology : CB, 30(10), 1934-1940 (2020-04-04)
Memory allows organisms to predict future events based on their prior sampling of the world. Rather than faithfully encoding each detail of related episodes, the brain is thought to incrementally construct probabilistic estimates of environmental statistics that are re-evaluated each

Articles

Nutrient Mixture F-12 Ham Formulation

Nutrient Mixture F-12 Ham Formulation

Nutrient Mixture F-12 Ham Formulation

Nutrient Mixture F-12 Ham Formulation

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service