Skip to Content
Merck
All Photos(3)

Key Documents

C9755

Sigma-Aldrich

Cytidine 5′-diphosphate sodium salt hydrate

from yeast, ≥95%

Synonym(s):

CDP sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N3O11P2
Molecular Weight:
403.18
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[Na].NC1=NC(=O)N(C=C1)C2OC(COP(O)(=O)OP(O)(O)=O)C(O)C2O

InChI

1S/C9H15N3O11P2.Na.H/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H2,10,11,15)(H2,16,17,18);;

InChI key

MAGSIBHRUIBONY-UHFFFAOYSA-N

Related Categories

General description

Cytidine 5′-diphosphate (CDP) is synthesized from cytidine monophosphate (CMP) or uridine monophosphate (UMP) by the transfer of phosphoryl group, catalyzed by the enzyme uridylate kinase (UMPK). CTP synthetase catalyzes the conversion of CTP to CDP intermediates like CDP-diacylglycerol, CDP-choline and CDP-ethanolamine for phospholipid biosynthesis.

Application

Cytidine 5′-diphosphate sodium salt hydrate has been used:
  • as a substrate in nucleoside diphosphatase (NDPase) enzyme histochemistry assay of gold fish eye sections
  • in ribonucleotide reductase assay
  • dCDP formation assay.

Biochem/physiol Actions

Cytidine 5′-diphosphate (CDP) is used as a substrate of nucleoside diphosphatase kinase to produce CTP for supporting DNA and RNA biosynthesis and of ribonucleotide reductase to product dCMP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Zhang et al.
Biochemistry, 50(25), 5615-5623 (2011-05-13)
Bacillus subtilis class Ib ribonucleotide reductase (RNR) catalyzes the conversion of nucleotides to deoxynucleotides, providing the building blocks for DNA replication and repair. It is composed of two proteins: α (NrdE) and β (NrdF). β contains the metallo-cofactor, essential for
Microglial cells in the retina of Carassius auratus: effects of optic nerve crush
Salvador-Silva M, et al.
The Journal of Comparative Neurology, 417(4), 431-447 (2000)
Purine and pyrimidine nucleotide synthesis and metabolism
The Arabidopsis Book / American Society of Plant Biologists, 1 (2002)
Ribonucleotide reductase requires subunit switching in hypoxia to maintain DNA replication
Foskolou IP, et al.
Molecular Cell, 66(2), 206-220 (2017)
CTP synthetase and its role in phospholipid synthesis in the yeast Saccharomyces cerevisiae
Chang YF and Carman GM
Progress in Lipid Research, 47(5), 333-339 (2008)

Protocols

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service