Skip to Content
Merck
All Photos(1)

Key Documents

C4654

Sigma-Aldrich

Cytidine

BioReagent, suitable for cell culture, powder, ≥99%

Synonym(s):

Cytosine β-D-riboside, Cytosine-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
Beilstein:
89173
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

210-220 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7-,8-/m1/s1

InChI key

UHDGCWIWMRVCDJ-XVFCMESISA-N

Gene Information

mouse ... Uck1(22245)

Looking for similar products? Visit Product Comparison Guide

General description

Cytidine is a nucleoside molecule. It is formed when cytosine is attached to a ribose ring through a β-N1-glycosidic bond.

Application

Cytidine has been used as a component to prepare ribonucleoside stock solution to assess its effects on the anaerobic growth of several Bacillus mojavensis strains.
Cytidine has been used as a non-essential aminoacid component of minimal essential medium (MEM) to analyse interspecies somatic cell nucleus transfer (iSCNT)-derived blastocysts. It has also been used as a supplement in the culture medium of HeLa cells, to study the effects of cytidine addition on rods and rings (RR) induced by glutamine deprivation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shramana Chatterjee et al.
Biochemical and biophysical research communications, 523(2), 348-353 (2019-12-24)
Protein cages have recently emerged as an extraordinary drug-delivery system due to its biocompatibility, biodegradability, low toxicity, ease to manipulate and engineer. We have reported earlier the formation and architecture of a do-decameric cage-like architecture of Vibrio cholerae acylphosphatase (VcAcP)
Beata Brozek-Pluska et al.
International journal of molecular sciences, 20(14) (2019-07-13)
Noninvasive Raman imaging of non-fixed and unstained human colon tissues based on vibrational properties of noncancerous and cancerous samples can effectively enable the differentiation between noncancerous and tumor tissues. This work aimed to evaluate the biochemical characteristics of colon cancer
Filippo M Cernilogar et al.
Nucleic acids research, 47(17), 9069-9086 (2019-07-28)
Pioneer transcription factors (PTF) can recognize their binding sites on nucleosomal DNA and trigger chromatin opening for recruitment of other non-pioneer transcription factors. However, critical properties of PTFs are still poorly understood, such as how these transcription factors selectively recognize
Interspecies somatic cell nucleus transfer with porcine oocytes as recipients: A novel bioassay system for assessing the competence of canine somatic cells to develop into embryos
Sugimura S, et al.
Theriogenology, 72(4), 549-559 (2009)
G P Gerrits et al.
Clinical chemistry, 34(7), 1439-1442 (1988-07-01)
Disturbances in the metabolism of purines and pyrimidines in neurologically affected patients can be reflected by aberrant concentrations of nucleosides and nucleobases in cerebrospinal fluid (CSF). However, normal values, especially for children at different ages, are lacking. We collected 1000

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service