Skip to Content
Merck

Skip To

C0985

Cytidine 3′,5′-cyclic mono­phos­phate sodium salt

≥98% (HPLC)

Synonym(s):

cCMP sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H11N3O7PNa
CAS Number:
Molecular Weight:
327.16
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
259-397-6
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

1 vial contains 50μmol or approximately 16mg

Quality Segment

assay

≥98% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@@H]3COP([O-])(=O)O[C@H]3[C@H]2O

InChI

1S/C9H12N3O7P.Na/c10-5-1-2-12(9(14)11-5)8-6(13)7-4(18-8)3-17-20(15,16)19-7;/h1-2,4,6-8,13H,3H2,(H,15,16)(H2,10,11,14);/q;+1/p-1/t4-,6-,7-,8-;/m1./s1

InChI key

RZAHVNCNTLFRHJ-IAIGYFSYSA-M

Biochem/physiol Actions

Produced by the hydrolysis of cytidine 5′-triphos­phate by the enzyme cytidylyl cyclase.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C9630SML1228D0260
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥97% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

description

1 vial contains 50μmol or approximately 16mg

description

-

description

contains 1μmol of powder (approx. 0.7mg)

description

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters