Skip to Content
Merck
All Photos(1)

Key Documents

C8988

Sigma-Aldrich

8-(4-Chlorophenylthio)-2′-O-methyladenosine 3′,5′-cyclic monophosphate monosodium hydrate

≥98% (HPLC)

Synonym(s):

8-pCPT-2′-O-Me-cAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N5O6ClPSNa · xH2O
CAS Number:
Molecular Weight:
507.82 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white

mp

235.5-237.5 °C (lit.)

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

O.[Na+].CO[C@@H]1[C@@H]2OP([O-])(=O)OC[C@H]2O[C@H]1n3c(Sc4ccc(Cl)cc4)nc5c(N)ncnc35

InChI

1S/C17H17ClN5O6PS.Na.H2O/c1-26-13-12-10(6-27-30(24,25)29-12)28-16(13)23-15-11(14(19)20-7-21-15)22-17(23)31-9-4-2-8(18)3-5-9;;/h2-5,7,10,12-13,16H,6H2,1H3,(H,24,25)(H2,19,20,21);;1H2/q;+1;/p-1/t10-,12-,13-,16-;;/m1../s1

InChI key

NVPSOXGMFVFZIS-IPBMGFFTSA-M

Biochem/physiol Actions

Analog of natural cAMP, potent and specific membrane-permeant activator of exchange factors directly activated by cAMP (Epac or cAMP-GEF), a new receptor for cyclic AMP

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang Tao et al.
Journal of oncology, 2022, 5483523-5483523 (2022-07-12)
Chemoresistance poses a great hindrance in the treatment of breast cancer (BC). Interestingly, exosome (Exo)-mediated transfer of long noncoding RNAs (lncRNAs) has been reported to regulate chemoresistance in diverse diseases. We herein investigate the potential role of lncRNA metastasis-associated lung
Mengye Li et al.
Clinical and experimental pharmacology & physiology, 44(6), 686-692 (2017-03-21)
Recent papers have attributed arrhythmic substrate in murine RyR2-P2328S hearts to reduced action potential (AP) conduction velocities (CV), reflecting acute functional inhibition and/or reduced expression of sodium channels. We explored for acute effects of direct exchange protein directly activated by
Ana-Caroline Raulin et al.
STAR protocols, 4(1), 101976-101976 (2023-01-05)
Cholesterol is a structural component of cell membranes. Most cells are incapable of its catabolism, and intracellular cholesterol accumulation is linked to several disorders including cardiovascular and neurodegenerative diseases. Cholesterol efflux, essential to its metabolism, is dependent on acceptors such
Yi-Fei Chen et al.
Respiratory research, 20(1), 285-285 (2019-12-20)
β2 receptor agonists induce airway smooth muscle relaxation by increasing intracellular cAMP production. PKA is the traditional downstream signaling pathway of cAMP. Exchange protein directly activated by cAMP (Epac) was identified as another important signaling molecule of cAMP recently. The
Andrzej J Chruscinski et al.
PloS one, 8(12), e82164-e82164 (2013-12-18)
β-adrenergic receptors (β-ARs) are model G-protein coupled receptors that mediate signal transduction in the sympathetic nervous system. Despite the widespread clinical use of agents that target β-ARs, the signaling pathways that operate downstream of β-AR stimulation have not yet been

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service