Skip to Content
Merck
All Photos(1)

Documents

F3917

Sigma-Aldrich

Forskolin

For use in molecular biology applications

Synonym(s):

7β-Acetoxy-8,13-epoxy-1α,6β,9α-trihydroxylabd-14-en-11-one, Coleonol, Colforsin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O7
CAS Number:
Molecular Weight:
410.50
Beilstein:
1692716
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

biological source

Coleus forskohlii

description

for activation of T cells

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

mol wt

410.50  g/mol

color

white to off-white

solubility

ethanol: soluble

suitability

suitable for inhibits interleukin 2 (IL-2) production in Jurkat cells

storage temp.

room temp

SMILES string

CC(=O)O[C@H]1[C@@H](O)[C@H]2C(C)(C)CC[C@H](O)[C@]2(C)[C@@]3(O)C(=O)C[C@@](C)(O[C@]13C)C=C

InChI

1S/C22H34O7/c1-8-19(5)11-14(25)22(27)20(6)13(24)9-10-18(3,4)16(20)15(26)17(28-12(2)23)21(22,7)29-19/h8,13,15-17,24,26-27H,1,9-11H2,2-7H3/t13-,15-,16-,17-,19-,20-,21+,22-/m0/s1

InChI key

OHCQJHSOBUTRHG-KGGHGJDLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Forskolin is a cell permeable diterpenoid isolated from Coleus forskohlii.

Biochem/physiol Actions

Cell-permeable diterpenoid that possesses anti-hypertensive, positive inotropic, and adenylyl cyclase activating properties. Many of its biological effects are due to its activation of adenylyl cyclase and the resulting increase in intracellular cAMP concentration. Forskolin effects calcium currents and inhibits MAP kinase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ramón-Cueto et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(10), 3803-3815 (1998-06-06)
The lack of axonal regeneration in the injured adult mammalian spinal cord leads to permanent functional impairment. To induce axonal regeneration in the transected adult rat spinal cord, we have used the axonal growth-promoting properties of adult olfactory bulb ensheathing
Jin Wei Chung et al.
Biology of reproduction, 86(3), 80-80 (2011-12-03)
The signaling molecule DHH, secreted by Sertoli cells, has essential regulatory functions in testicular differentiation. DHH is required for the differentiation of peritubular myoid cells that line the seminiferous cords and steroidogenic Leydig cells. The testicular cords in Dhh-null male
T Marchetti et al.
Journal of thrombosis and haemostasis : JTH, 12(6), 910-920 (2014-03-25)
Obstetric antiphospholipid syndrome (APS) is defined by pregnancy complications associated with antiphospholipid antibodies (aPL). The mechanisms of the pathogenic effects of aPL in pregnancy are poorly understood. Toll-like receptors (TLR) have been implicated previously in APS. The aims of our
Koji Yokoyama et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(1), 302-314 (2014-10-11)
Neonatal-onset multisystem inflammatory disease (NOMID) is a dominantly inherited autoinflammatory disease caused by NLRP3 mutations. NOMID pathophysiology is explained by the NLRP3 inflammasome, which produces interleukin-1β (IL-1β). However, epiphyseal overgrowth in NOMID is resistant to anti-IL-1 therapy and may therefore
Jeanne Ster et al.
The Journal of physiology, 587(1), 101-113 (2008-11-13)
Extensive work has shown that activation of the cAMP-dependent protein kinase A (PKA) is crucial for long-term depression (LTD) of synaptic transmission in the hippocampus, a phenomenon that is thought to be involved in memory formation. Here we studied the

Protocols

Learn how to cultivate similar-sized iPSC-derived colon organoids using Millicell® Microwell plates and perform a forskolin-induced swelling assay.

Learn how to cultivate similar-sized iPSC-derived colon organoids using Millicell® Microwell plates and perform a forskolin-induced swelling assay.

Learn how to cultivate similar-sized iPSC-derived colon organoids using Millicell® Microwell plates and perform a forskolin-induced swelling assay.

Learn how to cultivate similar-sized iPSC-derived colon organoids using Millicell® Microwell plates and perform a forskolin-induced swelling assay.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service