Skip to Content
Merck

C0406

κ-Casein from bovine milk

≥70% (PAGE), lyophilized powder

Synonym(s):

Kappa-Casein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
EC Number:
232-555-1
MDL number:
Form:
lyophilized powder
Assay:
≥70% (PAGE)
Biological source:
bovine milk
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Segment

assay

≥70% (PAGE)

form

lyophilized powder

technique(s)

isoelectric focusing (IEF): suitable

storage temp.

−20°C

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Gene Information

General description

Caseins constitute 80% of the total milk proteins in the cow. κ-Casein is one of the major allergens in cow milk. It is a glycosylated protein comprising galactose, galactosamine, and sialic acid. It exists as tri- or tetrasaccharides and occurs in different isoforms based on the number of oligosaccharides attached to it. κ-Casein has a hydrophobic N-terminus and a hydrophilic C-terminus region.

Application

κ-Casein from bovine milk has been used:
  • as a standard to quantify the casein concentration and to evaluate the ability of the bacterial strain to hydrolyze caseins released by HM Mozzarella
  • in LPS-depleted cow′s milk protein preparation for lymphoproliferation assay
  • as standard to quantify κ-casein concentration in skimmed milk samples by reversed-phase high-performance liquid chromatography (RP-HPLC)

Biochem/physiol Actions

κ-Casein from bovine milk is an immunoglobulin E (IgE)-binding epitope belonging to the casein phosphoprotein family. κ-Casein is important to the electrostatic and steric stabilization of casein micelles suspensions. It resists calcium precipitation and therefore, stabilizes other caseins. κ-Casein has been a target of research for creating low-phenylalanine milk through gene modification, which is important for patients suffering from metabolic diseases such as phenylketonuria (PKU).


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library